This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01225868_g1
See other SLC7A3 GE Assays ›
Gene Symbol:
Slc7a3
Gene Name:
solute carrier family 7 (cationic amino acid transporter, y+ system), member 3
Gene Aliases:
Atrc3, CAT-3, CAT3, SLC7A1, SLC7A2
Chromosome Location:
Chr.X: 101079204 - 101090452 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01225868_g1
Size
Availability Made To Order
Catalog # 4351372
Price 4,430.00
Your Price
Online offer:
4430.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Slc7a3
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
GenBank mRNA AK077593.1 - 9 - 10 1591 65
AK165931.1 - 9 - 10 1588 65
BC050195.1 - 9 - 10 1567 30010078 65
U70859.1 - 10 - 11 1725 65

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0X00002710.mm.1

Set Membership:

Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Panther Classification:

Molecular Function -

amino acid transporter

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0X00017040.mm.1 NM_007515.3 Refseq
Affymetrix Mouse Clariom S Array PSR0X00017040.mm.1 TC0X00002710.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0X00017042.mm.1 NM_007515.3 Refseq
Affymetrix Mouse Clariom S Array PSR0X00017042.mm.1 TC0X00002710.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-zjghb:80/100.66.79.197:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0