This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01225942_m1
See other OGT GE Assays ›
Gene Symbol:
Ogt
Gene Name:
O-linked N-acetylglucosamine (GlcNAc) transferase (UDP-N-acetylglucosamine:polypeptide-N-acetylglucosaminyl transferase)
Gene Aliases:
1110038P24Rik, 4831420N21Rik, AI115525, Ogtl
Chromosome Location:
Chr.X: 101640012 - 101684351 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01225942_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Ogt
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001290535.1 NP_001277464.1 18 - 19 2466 131
NM_139144.4 NP_631883.2 18 - 19 2663 131
GenBank mRNA AF363030.1 - 18 - 19 2428 131
AK045910.1 - 3 - 4 412 131
BC048239.1 - 14 - 15 2418 131
BC057319.1 - 18 - 19 2586 6830845 131

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0X00000981.mm.1

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Ogt

Panther Classification:

Molecular Function -

protein modifying enzyme

Gene Ontology Categories:

Function(s) Process(es)

negative regulation of protein phosphorylation
positive regulation of protein phosphorylation
glucosamine metabolic process
regulation of glycolytic process
protein O-linked glycosylation
apoptotic process
positive regulation of gene expression
negative regulation of peptidyl-threonine phosphorylation
chromatin modification
positive regulation of granulocyte differentiation
negative regulation of protein ubiquitination
response to insulin
circadian regulation of gene expression
negative regulation of peptidyl-serine phosphorylation
regulation of Rac protein signal transduction
positive regulation of catalytic activity
histone H4-K5 acetylation
histone H4-K8 acetylation
histone H4-K16 acetylation
positive regulation of cell size
positive regulation of proteolysis
positive regulation of transcription from RNA polymerase II promoter
regulation of insulin receptor signaling pathway
phosphatidylinositol-mediated signaling
intracellular distribution of mitochondria
rhythmic process
positive regulation of histone H3-K4 methylation
negative regulation of cell death
positive regulation of histone H3-K27 methylation
protein homotrimerization
protein heterotrimerization
cellular response to lipopolysaccharide
cellular response to retinoic acid
cellular response to glucose stimulus
histone H3-K4 trimethylation
negative regulation of protein targeting to membrane
regulation of gluconeogenesis involved in cellular glucose homeostasis
negative regulation of cellular response to hypoxia
positive regulation of protein localization to nucleus
positive regulation of reactive oxygen species biosynthetic process
catalytic activity
protein binding
phosphatidylinositol-3,4,5-trisphosphate binding
enzyme activator activity
N-acetyltransferase activity
transcription factor binding
lipid binding
protein N-acetylglucosaminyltransferase activity
transferase activity
transferase activity, transferring glycosyl groups
protein domain specific binding
peptide binding
histone acetyltransferase activity (H4-K5 specific)
histone acetyltransferase activity (H4-K8 specific)
histone acetyltransferase activity (H4-K16 specific)
monosaccharide binding
protein O-GlcNAc transferase activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0X00006604.mm.1 NM_139144.4 Refseq
Affymetrix Mouse Clariom S Array PSR0X00006604.mm.1 TC0X00000981.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-6b469b8bb7-lcddp:80/100.66.76.150:80.
git-commit: a334af76dff23450325448aedefe62379591458a
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.48.1-2026.04.16-1.0