This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01226660_m1
See other PPP1R12A GE Assays ›
Gene Symbol:
Ppp1r12a
Gene Name:
protein phosphatase 1, regulatory (inhibitor) subunit 12A
Gene Aliases:
1200015F06Rik, 5730577I22Rik, AA792106, AV099298, D10Ertd625e, Mypt1
Chromosome Location:
Chr.10: 108162065 - 108279646 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01226660_m1
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Ppp1r12a
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_027892.2 NP_082168.1 5 - 6 792 121
GenBank mRNA AK004785.1 - 5 - 6 792 121
BC125381.1 - 6 - 7 861 40129896 121
BC137630.1 - 6 - 7 861 8860649 121

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1000001266.mm.1

Set Membership:

Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons

Panther Classification:

Molecular Function -

phosphatase modulator

Gene Ontology Categories:

Function(s) Process(es)

protein dephosphorylation
mitotic nuclear division
signal transduction
regulation of cell adhesion
regulation of myosin-light-chain-phosphatase activity
negative regulation of catalytic activity
positive regulation of transcription from RNA polymerase II promoter
regulation of nucleocytoplasmic transport
centrosome organization
phosphoprotein phosphatase activity
enzyme inhibitor activity
protein binding
phosphatase regulator activity
protein kinase binding
14-3-3 protein binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1000009523.mm.1 NM_027892.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR1000009523.mm.1 NM_027892.2 Refseq
Affymetrix Mouse Clariom S Array PSR1000009523.mm.1 TC1000001266.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-rpjph:80/100.66.75.107:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0