This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01226748_m1
See other CLYBL GE Assays ›
Gene Symbol:
Clybl
Gene Name:
citrate lyase beta like
Gene Aliases:
0610033J05Rik, 2310014M14Rik, AI256068, Clb
Chromosome Location:
Chr.14: 122171283 - 122402234 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01226748_m1
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer:
435.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Clybl
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_029556.3 NP_083832.2 3 - 4 467 69
GenBank mRNA AF428254.1 - 3 - 4 440 69
AK002741.1 - 3 - 4 593 69
AK009345.1 - 3 - 4 455 69
BC023398.1 - 3 - 4 457 5064709 69

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the AK002741.1 transcripts.

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Clybl

Panther Classification:

Molecular Function -

lyase

Gene Ontology Categories:

Function(s) Process(es)

biological_process
molecular_function
catalytic activity
lyase activity
metal ion binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1400009961.mm.1 NM_029556.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR1400009961.mm.1 NM_029556.3 Refseq
Affymetrix Mouse Clariom S Array PSR1400009961.mm.1 TC1400001331.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-9dqhd:80/100.66.72.70:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline