This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01226773_g1
See other TWF2 GE Assays ›
Gene Symbol:
Twf2
Gene Name:
twinfilin, actin-binding protein, homolog 2 (Drosophila)
Gene Aliases:
A6-related, AU014993, Ptk9l, Ptk9r
Chromosome Location:
Chr.9: 106203108 - 106215387 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01226773_g1
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Twf2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_011876.3 NP_036006.1 3 - 4 436 77
GenBank mRNA AK002699.1 - 2 - 3 351 77
AK003997.1 - 2 - 3 331 77
AK084026.1 - 3 - 4 436 77
AK087422.1 - 2 - 3 351 77
AK155204.1 - 3 - 4 419 77
AK157886.1 - 3 - 4 381 77
AK166900.1 - 3 - 4 404 77
AK170441.1 - 3 - 4 384 77
AY267189.1 - 3 - 4 284 77
BC003338.1 - 3 - 4 383 3257230 77
Y17808.1 - 3 - 4 301 77

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0900001337.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Twf2

Panther Classification:

Molecular Function -

non-motor actin binding protein

Gene Ontology Categories:

Function(s) Process(es)

multicellular organism development
positive regulation of lamellipodium assembly
positive regulation of neuron projection development
cell projection organization
negative regulation of actin filament polymerization
regulation of microvillus length
regulation of actin cytoskeleton organization
intracellular signal transduction
sequestering of actin monomers
positive regulation of axon extension
barbed-end actin filament capping
cellular response to retinoic acid
cellular response to growth factor stimulus
actin binding
actin monomer binding
protein kinase C binding
ATP binding
phosphatidylinositol-4,5-bisphosphate binding
poly(A) RNA binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0900010433.mm.1 NM_011876.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR0900010433.mm.1 NM_011876.3 Refseq
Affymetrix Mouse Clariom S Array PSR0900010433.mm.1 TC0900001337.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-st8h5:80/100.66.79.144:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0