This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01226925_m1
See other DDX11 GE Assays ›
Gene Symbol:
Ddx11
Gene Name:
DEAD/H (Asp-Glu-Ala-Asp/His) box helicase 11
Gene Aliases:
4732462I11Rik, CHL1, CHLR1, KRG2, essa15a
Chromosome Location:
Chr.17: 66123501 - 66152174 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01226925_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Ddx11
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001003919.1 NP_001003919.1 5 - 6 730 129
GenBank mRNA AK049419.1 - 5 - 6 740 129
AK076357.1 - 5 - 6 759 129
AK152868.1 - 5 - 6 717 129
BC079656.1 - 5 - 6 730 6854039 129

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1700001084.mm.1

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Ddx11

Panther Classification:

Molecular Function -

DNA helicase

Gene Ontology Categories:

Function(s) Process(es)

nucleobase-containing compound metabolic process
cell cycle
sister chromatid cohesion
nucleotide binding
nucleic acid binding
DNA binding
double-stranded DNA binding
single-stranded DNA binding
RNA binding
ATP-dependent DNA helicase activity
helicase activity
ATP binding
ATP-dependent helicase activity
DNA-dependent ATPase activity
hydrolase activity
hydrolase activity, acting on acid anhydrides, in phosphorus-containing anhydrides
metal ion binding
iron-sulfur cluster binding
4 iron, 4 sulfur cluster binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1700010241.mm.1 NM_001003919.2 Refseq
Affymetrix Mouse Clariom S Array PSR1700010241.mm.1 TC1700001084.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1700010240.mm.1 NM_001003919.2 Refseq
Affymetrix Mouse Clariom S Array PSR1700010240.mm.1 TC1700001084.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-9dqhd:80/100.66.72.70:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline