This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01226965_m1
See other ABCG5 GE Assays ›
Gene Symbol:
Abcg5
Gene Name:
ATP-binding cassette, sub-family G (WHITE), member 5
Gene Aliases:
AW112016, cmp, sterolin-1, trac
Chromosome Location:
Chr.17: 84658234 - 84682923 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01226965_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Abcg5
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_031884.1 NP_114090.1 6 - 7 824 85
GenBank mRNA AF312713.2 - 6 - 7 912 85
AK149569.1 - 6 - 7 795 85
AY195872.1 - 6 - 7 912 85
AY195873.1 - 6 - 7 912 85
BC106766.2 - 6 - 7 801 40039484 85

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1700002655.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Transporter Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Abcg5

Panther Classification:

Molecular Function -

ATP-binding cassette (ABC) transporter primary active transporter transporter

Gene Ontology Categories:

Function(s) Process(es)

transport
excretion
negative regulation of intestinal phytosterol absorption
intestinal cholesterol absorption
cholesterol efflux
cholesterol homeostasis
negative regulation of intestinal cholesterol absorption
nucleotide binding
ATP binding
ATPase activity
cholesterol transporter activity
protein heterodimerization activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1700024072.mm.1 NM_031884.2 Refseq
Affymetrix Mouse Clariom S Array PSR1700024072.mm.1 TC1700002655.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1700024071.mm.1 NM_031884.2 Refseq
Affymetrix Mouse Clariom S Array PSR1700024071.mm.1 TC1700002655.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-kt9qp:80/100.66.76.204:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0