This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01227426_m1
See other SAFB GE Assays ›
Gene Symbol:
Safb
Gene Name:
scaffold attachment factor B
Gene Aliases:
3110021E02Rik, 5330423C17Rik, AU018122, D18386, E130307D12, HAP, HET, SAF-B1, SAFB1
Chromosome Location:
Chr.17: 56584760 - 56606294 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01227426_m1
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Safb
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001163300.1 NP_001156772.1 2 - 3 458 75
GenBank mRNA BC018200.1 - 2 - 3 434 75

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1700001010.mm.1

Set Membership:

Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

transcription, DNA-templated
regulation of transcription, DNA-templated
regulation of transcription from RNA polymerase II promoter
intracellular estrogen receptor signaling pathway
growth
hormone metabolic process
positive regulation of transcription from RNA polymerase II promoter
regulation of mRNA processing
nucleotide binding
core promoter binding
nucleic acid binding
DNA binding
chromatin binding
RNA binding
protein binding
protein kinase binding
sequence-specific DNA binding
poly(A) RNA binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1700009671.mm.1 NM_001163300.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1700009671.mm.1 NM_001163300.1 Refseq
Affymetrix Mouse Clariom S Array PSR1700009671.mm.1 TC1700001010.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1700009667.mm.1 NM_001163300.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1700009667.mm.1 NM_001163300.1 Refseq
Affymetrix Mouse Clariom S Array PSR1700009667.mm.1 TC1700001010.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1700009670.mm.1 NM_001163300.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1700009670.mm.1 NM_001163300.1 Refseq
Affymetrix Mouse Clariom S Array PSR1700009670.mm.1 TC1700001010.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1700009669.mm.1 NM_001163300.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1700009669.mm.1 NM_001163300.1 Refseq
Affymetrix Mouse Clariom S Array PSR1700009669.mm.1 TC1700001010.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1700009664.mm.1 NM_001163300.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1700009664.mm.1 NM_001163300.1 Refseq
Affymetrix Mouse Clariom S Array PSR1700009664.mm.1 TC1700001010.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1700009668.mm.1 NM_001163300.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1700009668.mm.1 NM_001163300.1 Refseq
Affymetrix Mouse Clariom S Array PSR1700009668.mm.1 TC1700001010.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1700009672.mm.1 NM_001163300.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1700009672.mm.1 NM_001163300.1 Refseq
Affymetrix Mouse Clariom S Array PSR1700009672.mm.1 TC1700001010.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-l4hk6:80/100.66.76.55:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0