This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01228024_m1
See other ARHGAP12 GE Assays ›
Gene Symbol:
Arhgap12
Gene Name:
Rho GTPase activating protein 12
Gene Aliases:
2810011M08Rik
Chromosome Location:
Chr.18: 6024448 - 6136098 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01228024_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Arhgap12
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001039692.1 NP_001034781.1 6 - 7 1564 109
NM_029277.2 NP_083553.2 6 - 7 1564 109
GenBank mRNA AK031648.1 - 6 - 7 1408 109
AK077063.1 - 6 - 7 1565 109
BC037608.1 - 6 - 7 1379 109
BC141184.1 - 6 - 7 1373 9055796 109
BC145506.1 - 6 - 7 1372 9054069 109

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1800000944.mm.1

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Arhgap12

Panther Classification:

Molecular Function -

GTPase-activating protein

Gene Ontology Categories:

Function(s) Process(es)

morphogenesis of an epithelial sheet
signal transduction
molecular_function
GTPase activator activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1800006906.mm.1 NM_001039692.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1800006906.mm.1 NM_001039692.1 Refseq
Affymetrix Mouse Clariom S Array PSR1800006906.mm.1 NM_029277.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR1800006906.mm.1 NM_029277.2 Refseq
Affymetrix Mouse Clariom S Array PSR1800006906.mm.1 TC1800000944.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-zpsr4:80/100.66.78.116:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0