This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01228030_m1
See other ZEB1 GE Assays ›
Gene Symbol:
Zeb1
Gene Name:
zinc finger E-box binding homeobox 1
Gene Aliases:
3110032K11Rik, AREB6, BZP, MEB1, Nil2, TCF-8, Tcf18, Tcf8, Tw, ZEB, Zfhep, Zfhx1a, Zfx1a, Zfx1ha, [delta]EF1
Chromosome Location:
Chr.18: 5591860 - 5775468 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01228030_m1
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer:
435.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Zeb1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_011546.3 NP_035676.1 5 - 6 775 63
GenBank mRNA AK140038.1 - 5 - 6 937 63
AK141437.1 - 1 - 2 105 63
BC090987.1 - 5 - 6 740 63
BC139768.1 - 5 - 6 744 40111224 63
BC139769.1 - 5 - 6 744 40111227 63
D76432.1 - 5 - 6 744 63
U26259.1 - 5 - 6 761 63

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1800000029.mm.1

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Zeb1

Panther Classification:

Molecular Function -

C2H2 zinc finger transcription factor

Gene Ontology Categories:

Function(s) Process(es)

negative regulation of transcription from RNA polymerase II promoter
transcription, DNA-templated
regulation of transcription, DNA-templated
transcription from RNA polymerase II promoter
pattern specification process
nervous system development
central nervous system development
negative regulation of cell proliferation
regulation of mesenchymal cell proliferation
regulation of transforming growth factor beta receptor signaling pathway
cell differentiation
negative regulation of epithelial cell differentiation
regulation of T cell differentiation in thymus
positive regulation of neuron differentiation
negative regulation of transcription, DNA-templated
positive regulation of transcription from RNA polymerase II promoter
animal organ development
embryonic camera-type eye morphogenesis
embryonic morphogenesis
embryonic skeletal system morphogenesis
semicircular canal morphogenesis
regulation of smooth muscle cell differentiation
cartilage development
cellular response to amino acid stimulus
cochlea morphogenesis
transcriptional repressor activity, RNA polymerase II transcription regulatory region sequence-specific binding
nucleic acid binding
DNA binding
chromatin binding
double-stranded DNA binding
transcription factor activity, sequence-specific DNA binding
protein binding
transcription factor binding
sequence-specific DNA binding
metal ion binding
E-box binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1800000240.mm.1 NM_011546.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR1800000240.mm.1 NM_011546.3 Refseq
Affymetrix Mouse Clariom S Array PSR1800000240.mm.1 TC1800000029.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1800000242.mm.1 NM_011546.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR1800000242.mm.1 NM_011546.3 Refseq
Affymetrix Mouse Clariom S Array PSR1800000242.mm.1 TC1800000029.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1800000241.mm.1 NM_011546.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR1800000241.mm.1 NM_011546.3 Refseq
Affymetrix Mouse Clariom S Array PSR1800000241.mm.1 TC1800000029.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1800000239.mm.1 NM_011546.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR1800000239.mm.1 NM_011546.3 Refseq
Affymetrix Mouse Clariom S Array PSR1800000239.mm.1 TC1800000029.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1800000243.mm.1 NM_011546.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR1800000243.mm.1 NM_011546.3 Refseq
Affymetrix Mouse Clariom S Array PSR1800000243.mm.1 TC1800000029.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-49tpj:80/100.66.77.222:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline