This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01228270_m1
See other REV1 GE Assays ›
Gene Symbol:
Rev1
Gene Name:
REV1 homolog (S. cerevisiae)
Gene Aliases:
1110027I23Rik, AU022044, Rev1l
Chromosome Location:
Chr.1: 38052786 - 38129919 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01228270_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Rev1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_019570.3 NP_062516.2 5 - 6 716 83
GenBank mRNA AB057418.1 - 4 - 5 505 83
AF179302.1 - 4 - 5 505 83
AK048838.1 - 5 - 6 721 83
BC058093.1 - 5 - 6 716 6827108 83

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0100002214.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Rev1

Panther Classification:

Molecular Function -

DNA metabolism protein nucleic acid metabolism protein

Gene Ontology Categories:

Function(s) Process(es)

DNA repair
cellular response to DNA damage stimulus
response to UV
error-prone translesion synthesis
DNA biosynthetic process
magnesium ion binding
DNA binding
damaged DNA binding
telomerase activity
RNA helicase activity
DNA-directed DNA polymerase activity
DNA primase activity
sulfate adenylyltransferase activity
RNA guanylyltransferase activity
tRNA guanylyltransferase activity
mannose-phosphate guanylyltransferase activity
transferase activity
nucleotidyltransferase activity
deoxycytidyl transferase activity
DNA/RNA helicase activity
CTP:2,3-di-O-geranylgeranyl-sn-glycero-1-phosphate cytidyltransferase activity
phospholactate guanylyltransferase activity
ATP:coenzyme F420 adenylyltransferase activity
metal ion binding
UDP-N-acetylgalactosamine diphosphorylase activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0100017884.mm.1 NM_019570.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR0100017884.mm.1 NM_019570.3 Refseq
Affymetrix Mouse Clariom S Array PSR0100017884.mm.1 TC0100002214.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-5thf6:80/100.66.79.225:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0