This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01228306_g1
See other MITD1 GE Assays ›
Gene Symbol:
Mitd1
Gene Name:
MIT, microtubule interacting and transport, domain containing 1
Gene Aliases:
1500032H18Rik
Chromosome Location:
Chr.1: 37878890 - 37890428 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01228306_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Mitd1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_026913.2 NP_081189.1 6 - 7 755 127
GenBank mRNA AK005342.1 - 6 - 7 689 127
AK171903.1 - 6 - 7 756 127
BC020137.1 - 751 3709157 127

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0100002210.mm.1

Set Membership:

Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Mus musculus 1700 Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

mitotic cytokinesis
cell separation after cytokinesis
transport
cell cycle
negative regulation of protein binding
cell division
protein domain specific binding
phosphatidylinositol binding
protein homodimerization activity

MicroArray Probes:

Agilent Affymetrix ABI Illumina

Array Name Probe IDs Targets Target Type
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P20156459 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P20156457 - -
Agilent Whole Mouse Genome Microarray 4x44K v2 A_66_P122110 AK171903.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_66_P122110 BC020137.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_66_P122110 NM_026913.2 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_66_P122110 AK005342.1 mouse_transcriptome
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 796868 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 88753 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 375331 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 375083 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 822055 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 551960 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 13328 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 250988 - -
Agilent SurePrint G3 Mouse CGH Microarray 4x180K 91472 - -
Agilent SurePrint G3 Mouse CGH Microarray 4x180K 120821 - -
Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P121115 - -
SurePrint G3 Mouse Gene Expression Microarray G4852A A_66_P122110 AK171903.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_66_P122110 BC020137.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_66_P122110 NM_026913.2 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_66_P122110 AK005342.1 mouse_transcriptome
Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P04067209 - -
Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P121115 - -
Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P00047217 - -
Whole Mouse Genome G4122A A_51_P303332 DV649848.1 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P303332 NM_001037918.3 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P303332 AK048005.1 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P303332 BC019519.1 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P303332 BC020137.1 mouse_transcriptome
Whole Mouse Genome G4122A A_52_P203833 AK005342.1 mouse_transcriptome
Whole Mouse Genome G4122A A_52_P203837 AK171903.1 mouse_transcriptome
Whole Mouse Genome G4122A A_52_P203837 BC020137.1 mouse_transcriptome
Whole Mouse Genome G4122A A_52_P203837 NM_026913.2 mouse_transcriptome
Whole Mouse Genome G4122A A_52_P203837 AK005342.1 mouse_transcriptome
Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P121115 - -
Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P00578507 - -
Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P00578511 - -
Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P00578504 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P20156457 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P20156459 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20156414 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20156370 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20156405 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20156397 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20156355 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20156447 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20156368 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20156347 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20156335 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20156412 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20156341 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20156351 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20156416 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20156364 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20156404 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20156385 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20156371 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20156393 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20156422 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20156345 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20156401 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20156340 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20156450 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20156374 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20156429 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20156407 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20156457 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20156410 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20156436 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20156338 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20156442 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20156349 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20156437 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20156455 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20156366 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20156358 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20156372 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20156377 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20156382 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20156336 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20156432 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20156409 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20156337 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20156381 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20156360 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20156384 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20156352 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20156395 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20156430 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20156389 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20156424 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20156417 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20156458 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20156453 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20156440 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20156411 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20156418 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20156415 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20156346 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20156362 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20156431 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P20156418 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P00669458 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P20156431 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P20156443 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P20156414 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P20156437 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P20156413 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P20156401 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P20156403 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P01103105 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P20156444 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P20156458 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P20156420 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P00139206 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P00397557 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P20156429 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P20156457 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P00243507 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P20156393 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P20156430 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P00139207 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P20156436 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P20156442 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P01056599 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P00660351 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P20156455 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P20156440 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P20156397 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P20156417 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P20156445 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P20156453 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P01056596 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P20156447 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P20156428 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P20156432 - -
Array Name Probe IDs Targets Target Type
Affymetrix Mouse Gene 1.0 ST Array 10354063 - -
Affymetrix Mouse Gene 1.0 ST Array 10354060 - -
Affymetrix Mouse Gene 1.0 ST Array 10354064 - -
Affymetrix Mouse Gene 1.0 ST Array 10354056 - -
Affymetrix Mouse Gene 1.0 ST Array 10354061 - -
Affymetrix Mouse Gene 1.0 ST Array 10354058 - -
Affymetrix Mouse Gene 1.0 ST Array 10354062 - -
Affymetrix Mouse Gene 1.0 ST Array 10354055 - -
Affymetrix Mouse Gene 1.0 ST Array 10354059 - -
Affymetrix Mouse Gene 1.0 ST Array 10354057 - -
Affymetrix HT MG-430 PM Array Plate 1439261_PM_x_at - -
Affymetrix HT MG-430 PM Array Plate 1426133_PM_a_at - -
Affymetrix Mouse Gene 1.1 ST Array 10354054 - -
Mouse Genome 430 2.0 Array 1439261_x_at - -
Mouse Genome 430 2.0 Array 1426133_a_at - -
Mouse Genome 430 2.0 Array 1439261_PM_x_at - -
Mouse Genome 430 2.0 Array 1426133_PM_a_at - -
Affymetrix GeneChip Mouse Genome 430 2.0 Array 69028_at - -
Affymetrix GeneChip Mouse Genome 430 2.0 Array 69028_PM_at - -
Affymetrix Mouse Clariom S Array PSR0100017803.mm.1 NM_026913.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0100017803.mm.1 NM_026913.2 Refseq
Affymetrix Mouse Clariom S Array PSR0100017803.mm.1 TC0100002210.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0100017801.mm.1 NM_026913.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0100017801.mm.1 NM_026913.2 Refseq
Affymetrix Mouse Clariom S Array PSR0100017801.mm.1 TC0100002210.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0100017802.mm.1 NM_026913.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0100017802.mm.1 NM_026913.2 Refseq
Affymetrix Mouse Clariom S Array PSR0100017802.mm.1 TC0100002210.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0100017798.mm.1 TC0100002210.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0100017800.mm.1 NM_026913.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0100017800.mm.1 NM_026913.2 Refseq
Affymetrix Mouse Clariom S Array PSR0100017800.mm.1 TC0100002210.mm.1 MTA TC
Mouse Expression Set 430 1439261_x_at - -
Mouse Expression Set 430 1426133_a_at - -
Mouse Expression Set 430 1439261_PM_x_at - -
Mouse Expression Set 430 1426133_PM_a_at - -
Mouse Genome 430A 2.0 Array 1426133_PM_a_at - -
Mouse Genome 430A 2.0 Array 1439261_PM_x_at - -
Mouse Genome 430A 2.0 Array 1426133_a_at - -
Mouse Genome 430A 2.0 Array 1439261_x_at - -
Affymetrix HT Mouse Genome 430A Array 1426133_a_at - -
Affymetrix HT Mouse Genome 430A Array 1426133_PM_a_at - -
Affymetrix HT Mouse Genome 430A Array 1439261_PM_x_at - -
Affymetrix HT Mouse Genome 430A Array 1439261_x_at - -
Array Name Probe IDs Targets Target Type
ABI Mouse Genome Survey Microarray 385915 - -
Array Name Probe IDs Targets Target Type
Illumina mouseRef-8 v1.1 expression beadchip ILMN_1217646 - -
Illumina mouse-6 v1.1 expression beadchip ILMN_1217646 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-8rxhs:80/100.66.78.116:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0