This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01230400_m1
See other UBA5 GE Assays ›
Gene Symbol:
Uba5
Gene Name:
ubiquitin-like modifier activating enzyme 5
Gene Aliases:
5730525G14Rik, AW240750, Ube1dc1
Chromosome Location:
Chr.9: 104046588 - 104063176 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01230400_m1
Size
Availability Made To Order
Catalog # 4351372
Price 4,430.00
Your Price
Online offer:
4430.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Uba5
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_025692.3 NP_079968.2 11 - 12 1307 72
GenBank mRNA AK017787.1 - 11 - 12 1307 72
AK077825.1 - 11 - 12 1288 72
AK151823.1 - 11 - 12 1264 72
AK152029.1 - 11 - 12 1302 72
AK168907.1 - 11 - 12 1301 72
AK178397.1 - 4 - 5 398 72

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 3' Most Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Uba5

Panther Classification:

Molecular Function -

ubiquitin-protein ligase

Gene Ontology Categories:

Function(s) Process(es)

regulation of intracellular estrogen receptor signaling pathway
response to endoplasmic reticulum stress
protein ufmylation
protein K69-linked ufmylation
nucleotide binding
ATP binding
small protein activating enzyme activity
metal ion binding
UFM1 activating enzyme activity

MicroArray Probes:

Agilent Affymetrix Illumina

Array Name Probe IDs Targets Target Type
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P26789499 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P26789498 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P26789500 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P26789501 - -
Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P403693 AK152029.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P403693 AK168907.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P403693 AK077825.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P403693 AK151823.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P403693 NM_025692.3 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P403693 AK152029.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P403693 AK168907.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P403693 AK077825.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P403693 AK151823.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P403693 NM_025692.3 mouse_transcriptome
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 891626 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 78089 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 371308 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 45163 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 18322 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 668151 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 43512 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 628161 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 280895 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 158199 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 782660 - -
Agilent SurePrint G3 Mouse CGH Microarray 4x180K 28510 - -
Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P133543 - -
Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P172673 - -
SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P403693 AK152029.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P403693 AK168907.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P403693 AK077825.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P403693 AK151823.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P403693 NM_025692.3 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P403693 AK152029.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P403693 AK168907.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P403693 AK077825.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P403693 AK151823.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P403693 NM_025692.3 mouse_transcriptome
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26789471 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26789470 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26789487 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26789485 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26789476 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26789496 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26789465 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26789479 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26789467 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26789500 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26789463 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26789501 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26789469 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26789503 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26789491 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26789493 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26789474 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26789462 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26789482 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26789488 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26789489 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26789464 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26789484 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26789504 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26789480 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26789477 - -
Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P06908875 - -
Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P133543 - -
Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P172673 - -
Whole Mouse Genome G4122A A_51_P386046 - -
Whole Mouse Genome G4122A A_51_P403693 AK152029.1 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P403693 AK168907.1 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P403693 AK077825.1 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P403693 AK151823.1 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P403693 NM_025692.3 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P403693 AK152029.1 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P403693 AK168907.1 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P403693 AK077825.1 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P403693 AK151823.1 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P403693 NM_025692.3 mouse_transcriptome
Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P124047 - -
Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P172673 - -
Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P133543 - -
Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P07730178 - -
Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P07617954 - -
Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P07617955 - -
Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P07617956 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P26789500 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P26789501 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P26789499 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P26789498 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26789462 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26789500 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26789487 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26789464 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26789406 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26789459 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26789461 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26789493 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26789414 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26789428 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26789496 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26789460 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26789504 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26789450 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26789458 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26789469 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26789398 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26789401 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26789448 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26789484 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26789503 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26789413 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26789411 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26789420 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26789441 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26789402 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26789435 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26789501 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26789498 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26789455 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26789434 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26789431 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26789489 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26789399 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26789409 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26789439 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26789437 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26789465 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26789415 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26789421 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26789474 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26789423 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26789408 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26789491 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26789463 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26789404 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26789405 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26789485 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26789480 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26789429 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26789425 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26789410 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26789467 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26789444 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26789451 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26789477 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26789482 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26789470 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26789426 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26789445 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P26789472 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P26789446 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P26789477 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P26789451 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P26789463 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P26789453 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P26789448 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P26789406 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P26789470 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P08078604 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P26789465 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P26789482 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P26789469 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P26789405 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P26789398 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P07764206 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P26789496 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P07764839 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P26789450 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P26789484 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P07575423 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P26789404 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P26789462 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P07764201 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P26789460 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P26789480 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P26789471 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P26789455 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P07730179 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P26789401 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P26789498 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P26789476 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P26789399 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P26789489 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P07764834 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P26789485 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P26789464 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P26789503 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P26789395 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P26789479 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P26789491 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P07616578 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P07930560 - -
Array Name Probe IDs Targets Target Type
Affymetrix Mouse Gene 1.0 ST Array 10596212 - -
Affymetrix Mouse Gene 1.0 ST Array 10596214 - -
Affymetrix Mouse Gene 1.0 ST Array 10596220 - -
Affymetrix Mouse Gene 1.0 ST Array 10596209 - -
Affymetrix Mouse Gene 1.0 ST Array 10596208 - -
Affymetrix Mouse Gene 1.0 ST Array 10596217 - -
Affymetrix Mouse Gene 1.0 ST Array 10596219 - -
Affymetrix Mouse Gene 1.0 ST Array 10596221 - -
Affymetrix Mouse Gene 1.0 ST Array 10596213 - -
Affymetrix Mouse Gene 1.0 ST Array 10596210 - -
Affymetrix Mouse Gene 1.0 ST Array 10596218 - -
Affymetrix Mouse Gene 1.0 ST Array 10596216 - -
Affymetrix Mouse Gene 1.0 ST Array 10596215 - -
Affymetrix Mouse Gene 1.0 ST Array 10596211 - -
Affymetrix HT MG-430 PM Array Plate 1435247_PM_at - -
Affymetrix HT MG-430 PM Array Plate 1451973_PM_at - -
Affymetrix Mouse Gene 1.1 ST Array 10596207 - -
Mouse Genome 430 2.0 Array 1451973_at - -
Mouse Genome 430 2.0 Array 1435247_at - -
Mouse Genome 430 2.0 Array 1451973_PM_at - -
Mouse Genome 430 2.0 Array 1435247_PM_at - -
Affymetrix GeneChip Mouse Genome 430 2.0 Array 66663_at - -
Affymetrix GeneChip Mouse Genome 430 2.0 Array 66663_PM_at - -
Mouse Expression Set 430 1451973_at - -
Mouse Expression Set 430 1451973_PM_at - -
Mouse Expression Set 430 1435247_PM_at - -
Mouse Expression Set 430 1435247_at - -
Murine Genome U74v2 Set 102813_f_at - -
Murine Genome U74v2 Set 102813_PM_f_at - -
Murine Genome U74v2 Set 102812_i_at - -
Murine Genome U74v2 Set 102812_PM_i_at - -
Mouse Genome 430A 2.0 Array 1451973_at - -
Mouse Genome 430A 2.0 Array 1451973_PM_at - -
Affymetrix HT Mouse Genome 430A Array 1451973_at - -
Affymetrix HT Mouse Genome 430A Array 1451973_PM_at - -
Affymetrix HT Mouse Genome 430B Array 1435247_at - -
Affymetrix HT Mouse Genome 430B Array 1435247_PM_at - -
Array Name Probe IDs Targets Target Type
MouseWG-6 v2 Expression BeadChip 110021 - -
Illumina mouse-6 v1.0 expression beadchip 110021 - -
Illumina mouseRef-8 v1.0 expression beadchip 0000110021 - -
Illumina mouseRef-8 v1.1 expression beadchip ILMN_1216388 - -
Illumina mouse-6 v1.1 expression beadchip ILMN_1216388 - -
mouse-6 v1.1 (Illumina) scl35421.10.1_30-S - -
Illumina Sentrix mouse-6.v1 bead array 110021 - -
Illumina Sentrix MouseRef-8 Expression BeadChip scl35421.10.1_30-S - -
Illumina Sentrix Mouse-6 Expression BeadChip (2) 110021 - -
MouseRef-8 v2 Expression BeadChip ILMN_1216388 - -
Illumina MouseWG-6 v2.0 expression beadchip ILMN_1216388 - -
Illumina Sentrix Mouse-6 Expression BeadChip (3) scl35421.10.1_30-S - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-zjghb:80/100.66.79.197:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0