This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01230401_m1
See other UBA5 GE Assays ›
Gene Symbol:
Uba5
Gene Name:
ubiquitin-like modifier activating enzyme 5
Gene Aliases:
5730525G14Rik, AW240750, Ube1dc1
Chromosome Location:
Chr.9: 104046588 - 104063176 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01230401_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Uba5
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_025692.3 NP_079968.2 1 - 2 326 59
GenBank mRNA AK017787.1 - 1 - 2 326 59
AK077825.1 - 1 - 2 308 59
AK151823.1 - 1 - 2 283 59
AK152029.1 - 1 - 2 321 59
AK168907.1 - 1 - 2 320 59
AK206146.1 - 1 - 2 43 59
BC019764.1 - 1 - 2 203 3989119 59
BY768723.1 - 1 - 2 327 59

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus 1700 Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Uba5

Panther Classification:

Molecular Function -

ubiquitin-protein ligase

Gene Ontology Categories:

Function(s) Process(es)

regulation of intracellular estrogen receptor signaling pathway
response to endoplasmic reticulum stress
protein ufmylation
protein K69-linked ufmylation
nucleotide binding
ATP binding
small protein activating enzyme activity
metal ion binding
UFM1 activating enzyme activity

MicroArray Probes:

ABI

Array Name Probe IDs Targets Target Type
ABI Mouse Genome Survey Microarray 686402 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-2xklz:80/100.66.76.139:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0