This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01230403_m1
See other UBA5 GE Assays ›
Gene Symbol:
Uba5
Gene Name:
ubiquitin-like modifier activating enzyme 5
Gene Aliases:
5730525G14Rik, AW240750, Ube1dc1
Chromosome Location:
Chr.9: 104046588 - 104063176 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01230403_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Uba5
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_025692.3 NP_079968.2 4 - 5 574 141
GenBank mRNA AK077825.1 - 4 - 5 555 141
AK151823.1 - 4 - 5 531 141
AK152029.1 - 4 - 5 569 141
AK168907.1 - 4 - 5 568 141
BC019764.1 - 4 - 5 451 3989119 141

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0900002981.mm.1

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Uba5

Panther Classification:

Molecular Function -

ubiquitin-protein ligase

Gene Ontology Categories:

Function(s) Process(es)

regulation of intracellular estrogen receptor signaling pathway
response to endoplasmic reticulum stress
protein ufmylation
protein K69-linked ufmylation
nucleotide binding
ATP binding
small protein activating enzyme activity
metal ion binding
UFM1 activating enzyme activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0900024519.mm.1 NM_025692.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR0900024519.mm.1 NM_025692.3 Refseq
Affymetrix Mouse Clariom S Array PSR0900024519.mm.1 TC0900002981.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-zpsr4:80/100.66.78.116:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0