This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01230631_g1
See other RBM6 GE Assays ›
Gene Symbol:
Rbm6
Gene Name:
RNA binding motif protein 6
Gene Aliases:
4930506F14Rik, Def-3, NY-LU-12, g16, mKIAA4015
Chromosome Location:
Chr.9: 107773559 - 107872819 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01230631_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Rbm6
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_011251.3 NP_035381.2 18 - 19 3238 132
NM_029169.3 NP_083445.1 18 - 19 3509 132
GenBank mRNA AJ006486.2 - 18 - 19 3114 132
AK077899.1 - 18 - 19 3417 133
AK145612.1 - 18 - 19 3398 132
AK164432.1 - 13 - 14 1595 132
AK220446.1 - 18 - 19 3384 132

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Rbm6

Gene Ontology Categories:

Process(es)

poly(A) RNA binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-cjlz8:80/100.66.74.50:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0