This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01230937_g1
See other POLR2G GE Assays ›
Gene Symbol:
Polr2g
Gene Name:
polymerase (RNA) II (DNA directed) polypeptide G
Gene Aliases:
2410046K11Rik, A230108L04Rik, C76415, RBP7, Rpo2-7l
Chromosome Location:
Chr.19: 8793129 - 8798557 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01230937_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Polr2g
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_026329.2 NP_080605.1 4 - 5 419 98
GenBank mRNA AK010687.1 - 4 - 5 388 98
AK011627.1 - 4 - 5 420 98
AK013134.1 - 4 - 5 390 98
AK017473.1 - 4 - 5 398 98
BC005580.1 - 4 - 5 438 3968561 98
BC055278.1 - 4 - 5 369 5699685 98

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1900001070.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Polr2g

Panther Classification:

Molecular Function -

DNA-directed RNA polymerase

Gene Ontology Categories:

Function(s) Process(es)

nuclear-transcribed mRNA catabolic process, exonucleolytic
transcription, DNA-templated
transcription from RNA polymerase II promoter
transcription initiation from RNA polymerase II promoter
positive regulation of translational initiation
positive regulation of nuclear-transcribed mRNA poly(A) tail shortening
nucleic acid binding
single-stranded DNA binding
RNA binding
single-stranded RNA binding
DNA-directed RNA polymerase activity
transferase activity
nucleotidyltransferase activity
translation initiation factor binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1900010029.mm.1 NM_026329.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR1900010029.mm.1 NM_026329.2 Refseq
Affymetrix Mouse Clariom S Array PSR1900010029.mm.1 TC1900001070.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-8cb8z:80/100.66.72.77:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0