This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01230944_g1
See other CREM GE Assays ›
Gene Symbol:
Crem
Gene Name:
cAMP responsive element modulator
Gene Aliases:
ICER, ICERI
Chromosome Location:
Chr.18: 3266354 - 3340355 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01230944_g1
Size
Availability Inventoried
Catalog # 4331182
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Crem
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001110850.2 NP_001104320.1 3 - 4 307 124
NM_001110851.1 NP_001104321.1 4 - 5 566 124
NM_001110852.1 NP_001104322.1 3 - 4 419 124
NM_001110853.2 NP_001104323.1 3 - 4 345 124
NM_001110856.2 NP_001104326.1 4 - 5 454 124
NM_001110857.2 NP_001104327.1 3 - 4 307 124
NM_001110858.2 NP_001104328.1 3 - 4 307 124
NM_001110859.2 NP_001104329.1 4 - 5 454 124
NM_001271503.1 NP_001258432.1 4 - 5 454 124
NM_001271504.1 NP_001258433.1 4 - 5 454 124
NM_001271505.1 NP_001258434.1 3 - 4 307 124
NM_001271506.1 NP_001258435.1 4 - 5 454 124
NM_013498.3 NP_038526.2 3 - 4 307 124
NR_073193.1 5 - 6 550 124
GenBank mRNA AI461618.1 - 4 - 5 416 124
AK009841.1 - 3 - 4 339 124
AK010806.1 - 3 - 4 420 124
AK015641.1 - 3 - 4 346 124
AK016156.1 - 3 - 4 332 124
AK076797.1 - 3 - 4 268 124
AK161366.1 - 4 - 5 500 124
AY738715.1 - 3 - 4 370 124
AY738716.1 - 2 - 3 223 124
AY738717.1 - 3 - 4 370 124
AY738718.1 - 3 - 4 370 124
AY738719.1 - 2 - 3 223 124
AY738720.1 - 4 - 5 466 124
AY738721.1 - 3 - 4 370 124
BC034856.1 - 2 - 3 216 124
M60285.1 - 3 - 4 235 124
S64590.1 - 3 - 4 352 124

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Mus musculus 1700 Inventoried Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Crem

Gene Ontology Categories:

Function(s) Process(es)

transcription, DNA-templated
regulation of transcription, DNA-templated
glycosphingolipid metabolic process
spermatogenesis
circadian regulation of gene expression
negative regulation of transcription, DNA-templated
positive regulation of transcription from RNA polymerase II promoter
rhythmic process
response to cAMP
core promoter sequence-specific DNA binding
DNA binding
transcription factor activity, sequence-specific DNA binding
protein binding
sequence-specific DNA binding

MicroArray Probes:

ABI

Array Name Probe IDs Targets Target Type
ABI Mouse Genome Survey Microarray 927419 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-9w5rb:80/100.66.76.55:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0