This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01231349_m1
See other PELI3 GE Assays ›
Gene Symbol:
Peli3
Gene Name:
pellino 3
Gene Aliases:
6030441F14Rik, A930011L17, BC028931
Chromosome Location:
Chr.19: 4929717 - 4943127 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01231349_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Peli3
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_172835.3 NP_766423.3 6 - 7 922 65
GenBank mRNA AK044418.1 - 6 - 7 869 65
AK135575.1 - 5 - 6 770 65
BC028931.1 - 6 - 7 880 5037308 65

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1900000943.mm.1

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Mus musculus 1700 Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Peli3

Panther Classification:

Molecular Function -

ubiquitin-protein ligase protein modifying enzyme

Gene Ontology Categories:

Function(s) Process(es)

Toll signaling pathway
negative regulation of tumor necrosis factor-mediated signaling pathway
protein ubiquitination
negative regulation of protein ubiquitination
negative regulation of type I interferon production
negative regulation of Toll signaling pathway
defense response to Gram-negative bacterium
regulation of nucleotide-binding oligomerization domain containing 1 signaling pathway
positive regulation of nucleotide-binding oligomerization domain containing 2 signaling pathway
protein K63-linked ubiquitination
negative regulation of extrinsic apoptotic signaling pathway
ubiquitin-protein transferase activity
protein binding
UDP-N-acetylmuramoylalanyl-D-glutamyl-2,6-diaminopimelate-D-alanyl-D-alanine ligase activity
ligase activity
ribosomal S6-glutamic acid ligase activity
coenzyme F420-0 gamma-glutamyl ligase activity
coenzyme F420-2 alpha-glutamyl ligase activity
ubiquitin protein ligase activity
protein-glycine ligase activity
protein-glycine ligase activity, initiating
protein-glycine ligase activity, elongating
tubulin-glycine ligase activity
protein-glutamic acid ligase activity
tubulin-glutamic acid ligase activity

MicroArray Probes:

Agilent Affymetrix ABI Illumina

Array Name Probe IDs Targets Target Type
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31926008 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31926009 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31926005 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31926004 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31926006 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31926003 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31926007 - -
Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P266695 BY452323.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P266695 AK044418.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P266695 AK135575.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P266695 BC028931.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P266695 NM_172835.3 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P266695 BY452323.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P266695 AK044418.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P266695 AK135575.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P266695 BC028931.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P266695 NM_172835.3 mouse_transcriptome
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 512062 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 119451 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 325889 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 14715 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 845820 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 637544 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 763199 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 368854 - -
Agilent SurePrint G3 Mouse CGH Microarray 4x180K 139213 - -
Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P174673 - -
Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P156539 - -
SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P266695 BY452323.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P266695 AK044418.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P266695 AK135575.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P266695 BC028931.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P266695 NM_172835.3 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P266695 BY452323.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P266695 AK044418.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P266695 AK135575.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P266695 BC028931.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P266695 NM_172835.3 mouse_transcriptome
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31926018 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31926019 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31925990 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31926027 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31925934 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31926002 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31926023 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31925993 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31926015 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31926013 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31926008 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31926025 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31926032 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31926009 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31925999 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31925991 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31926029 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31925931 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31926010 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31926001 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31926030 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31925937 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31926005 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31925938 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31925935 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31925998 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31925994 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31926007 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31925940 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31925942 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31926014 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31926021 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31925933 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31926012 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31925932 - -
Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P09092031 - -
Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P174673 - -
Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P156539 - -
Whole Mouse Genome G4122A A_51_P266695 BY452323.1 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P266695 AK044418.1 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P266695 AK135575.1 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P266695 BC028931.1 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P266695 NM_172835.3 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P266695 BY452323.1 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P266695 AK044418.1 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P266695 AK135575.1 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P266695 BC028931.1 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P266695 NM_172835.3 mouse_transcriptome
Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P139371 - -
Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P156539 - -
Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P174673 - -
Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P14342927 - -
Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P14342937 - -
Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P14342943 - -
Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P14342941 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31926005 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31926004 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31926007 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31926009 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31926006 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31926003 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31926008 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31925944 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31925987 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31925867 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31925919 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31925934 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31925930 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31925938 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31926001 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31925959 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31925927 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31925916 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31925999 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31925993 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31925909 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31925928 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31926007 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31926021 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31925881 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31925882 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31926002 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31926010 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31926015 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31925894 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31926039 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31925912 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31926005 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31925969 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31925871 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31926019 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31925961 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31925878 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31926030 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31926023 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31925896 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31925917 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31925892 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31925937 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31925986 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31925947 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31925945 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31925964 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31925866 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31925957 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31926008 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31925899 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31925865 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31925908 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31926014 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31925931 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31926025 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31925879 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31925994 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31925956 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31925923 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31925890 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31925907 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31925889 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31925963 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31925915 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31925932 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31925926 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31925974 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31925975 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31925873 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31925905 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31925921 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31925898 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31925888 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31925875 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31925973 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31925984 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31925900 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31925902 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31925913 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31926038 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31925951 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31926018 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31925940 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31925886 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31925904 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31925953 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P31925934 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P31925956 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P31926023 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P31926021 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P31925989 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P31925945 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P31925966 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P31925987 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P14498985 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P31925932 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P14199670 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P31925968 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P14265461 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P31925969 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P14265495 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P31926005 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P14207308 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P31926018 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P14490554 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P14342928 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P14342936 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P31925961 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P14344271 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P31925973 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P31926028 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P31926025 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P31925947 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P31925993 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P31925999 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P31925963 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P31926016 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P31925990 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P14413225 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P14342925 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P31925940 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P31925937 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P31926010 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P14161874 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P31925965 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P31926019 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P14498988 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P31926013 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P31925986 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P31926030 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P14342945 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P31925974 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P31925931 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P31925951 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P14328810 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P14342927 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P14488620 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P31925964 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P31925953 - -
Array Name Probe IDs Targets Target Type
Affymetrix Mouse Gene 1.0 ST Array 10464897 - -
Affymetrix Mouse Gene 1.0 ST Array 10464900 - -
Affymetrix Mouse Gene 1.0 ST Array 10464899 - -
Affymetrix Mouse Gene 1.0 ST Array 10464898 - -
Affymetrix Mouse Gene 1.0 ST Array 10464903 - -
Affymetrix Mouse Gene 1.0 ST Array 10464901 - -
Affymetrix Mouse Gene 1.0 ST Array 10464904 - -
Affymetrix Mouse Gene 1.1 ST Array 10464896 - -
Affymetrix Mouse Clariom S Array PSR1900008331.mm.1 NM_172835.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR1900008331.mm.1 NM_172835.3 Refseq
Affymetrix Mouse Clariom S Array PSR1900008331.mm.1 TC1900000943.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1900008332.mm.1 NM_172835.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR1900008332.mm.1 NM_172835.3 Refseq
Affymetrix Mouse Clariom S Array PSR1900008332.mm.1 TC1900000943.mm.1 MTA TC
Array Name Probe IDs Targets Target Type
ABI Mouse Genome Survey Microarray 333982 - -
Array Name Probe IDs Targets Target Type
Illumina mouse-6 v1.0 expression beadchip 6770438 - -
Illumina mouse-6 v1.0 expression beadchip 3360685 - -
Illumina mouseRef-8 v1.0 expression beadchip 0006770438 - -
Illumina mouseRef-8 v1.0 expression beadchip 0003360685 - -
Illumina mouseRef-8 v1.1 expression beadchip ILMN_1239882 - -
Illumina mouseRef-8 v1.1 expression beadchip ILMN_2619678 - -
Illumina mouse-6 v1.1 expression beadchip ILMN_2619678 - -
Illumina mouse-6 v1.1 expression beadchip ILMN_1239882 - -
mouse-6 v1.1 (Illumina) scl00240518.1_93-S - -
mouse-6 v1.1 (Illumina) scl53481.7.1_229-S - -
Illumina Sentrix mouse-6.v1 bead array 6770438 - -
Illumina Sentrix mouse-6.v1 bead array 3360685 - -
Illumina Sentrix MouseRef-8 Expression BeadChip scl53481.7.1_229-S - -
Illumina Sentrix MouseRef-8 Expression BeadChip scl00240518.1_93-S - -
Illumina Sentrix Mouse-6 Expression BeadChip (2) 3360685 - -
Illumina Sentrix Mouse-6 Expression BeadChip (2) 6770438 - -
MouseRef-8 v2 Expression BeadChip ILMN_2619678 - -
MouseRef-8 v2 Expression BeadChip ILMN_1239882 - -
Illumina MouseWG-6 v2.0 expression beadchip ILMN_1239882 - -
Illumina MouseWG-6 v2.0 expression beadchip ILMN_2619678 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-2xklz:80/100.66.76.139:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0