This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01231368_m1
See other YIF1A GE Assays ›
Gene Symbol:
Yif1a
Gene Name:
Yip1 interacting factor homolog A (S. cerevisiae)
Gene Aliases:
5330422J23Rik, 54TM, Yif1, Yif1p
Chromosome Location:
Chr.19: 5088538 - 5092879 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01231368_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Yif1a
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_026553.4 NP_080829.1 4 - 5 607 60
GenBank mRNA AK077329.1 - 4 - 5 593 60
AK160409.1 - 4 - 5 560 60
BC011117.1 - 4 - 5 549 4190535 60

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1900000056.mm.1

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Yif1a

Gene Ontology Categories:

Function(s) Process(es)

transport
biological_process
protein transport
vesicle-mediated transport
molecular_function

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1900000734.mm.1 NM_026553.4.1 Refseq
Affymetrix Mouse Clariom S Array PSR1900000734.mm.1 NM_026553.4 Refseq
Affymetrix Mouse Clariom S Array PSR1900000734.mm.1 TC1900000056.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-zpsr4:80/100.66.78.116:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0