This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01231749_m1
See other GPR137 GE Assays ›
Gene Symbol:
Gpr137
Gene Name:
G protein-coupled receptor 137
Gene Aliases:
AI428855
Chromosome Location:
Chr.19: 6938062 - 6942450 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01231749_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Gpr137
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001177362.1 NP_001170833.1 3 - 4 577 58
GenBank mRNA AK170308.1 - 3 - 4 579 58

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1900001023.mm.1

Set Membership:

Amplicon length less than or equal to 70 G-protein coupled receptor Mus musculus Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

biological_process
molecular_function

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1900009504.mm.1 NM_001177360.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1900009504.mm.1 NM_001177360.1 Refseq
Affymetrix Mouse Clariom S Array PSR1900009504.mm.1 NM_001177362.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1900009504.mm.1 NM_001177362.1 Refseq
Affymetrix Mouse Clariom S Array PSR1900009504.mm.1 NM_207220.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR1900009504.mm.1 NM_207220.2 Refseq
Affymetrix Mouse Clariom S Array PSR1900009504.mm.1 TC1900001023.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1900009508.mm.1 NM_001177362.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1900009508.mm.1 NM_001177362.1 Refseq
Affymetrix Mouse Clariom S Array PSR1900009508.mm.1 TC1900001023.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1900009507.mm.1 NM_001177362.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1900009507.mm.1 NM_001177362.1 Refseq
Affymetrix Mouse Clariom S Array PSR1900009507.mm.1 TC1900001023.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1900009509.mm.1 NM_001177362.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1900009509.mm.1 NM_001177362.1 Refseq
Affymetrix Mouse Clariom S Array PSR1900009509.mm.1 TC1900001023.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1900009498.mm.1 NM_001177360.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1900009498.mm.1 NM_001177360.1 Refseq
Affymetrix Mouse Clariom S Array PSR1900009498.mm.1 NM_001177361.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1900009498.mm.1 NM_001177361.1 Refseq
Affymetrix Mouse Clariom S Array PSR1900009498.mm.1 NM_001177362.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1900009498.mm.1 NM_001177362.1 Refseq
Affymetrix Mouse Clariom S Array PSR1900009498.mm.1 NM_207220.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR1900009498.mm.1 NM_207220.2 Refseq
Affymetrix Mouse Clariom S Array PSR1900009498.mm.1 TC1900001023.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1900009506.mm.1 NM_001177360.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1900009506.mm.1 NM_001177360.1 Refseq
Affymetrix Mouse Clariom S Array PSR1900009506.mm.1 NM_001177362.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1900009506.mm.1 NM_001177362.1 Refseq
Affymetrix Mouse Clariom S Array PSR1900009506.mm.1 NM_207220.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR1900009506.mm.1 NM_207220.2 Refseq
Affymetrix Mouse Clariom S Array PSR1900009506.mm.1 TC1900001023.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-8cb8z:80/100.66.72.77:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0