This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01232095_m1
See other ATRIP GE Assays ›
Gene Symbol:
Atrip
Gene Name:
ATR interacting protein
Gene Aliases:
6620401K05Rik
Chromosome Location:
Chr.9: 109059747 - 109075580 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01232095_m1
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Atrip
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_172774.3 NP_766362.2 3 - 4 673 76
GenBank mRNA AK031257.1 - 3 - 4 663 76
AK171423.1 - 3 - 4 628 76
AK189287.1 - 2 - 3 290 76
BC078458.1 - 3 - 4 620 6832436 76

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0900003073.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Atrip

Gene Ontology Categories:

Function(s) Process(es)

DNA repair
cellular response to DNA damage stimulus
protein binding
K63-linked polyubiquitin binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0900025906.mm.1 NM_172774.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR0900025906.mm.1 NM_172774.3 Refseq
Affymetrix Mouse Clariom S Array PSR0900025906.mm.1 TC0900003073.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-55658555d6-9rjdb:80/100.66.79.224:80.
git-commit: ec8e7df5f8fec8d765bd419205f1b5046a016d37
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.41.0-Offline