This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01232361_m1
See other TWSG1 GE Assays ›
Gene Symbol:
Twsg1
Gene Name:
twisted gastrulation homolog 1 (Drosophila)
Gene Aliases:
1810013J15Rik, 9030422N06Rik, AW552143, D17Ertd403e, Tsg, Twg
Chromosome Location:
Chr.17: 65923065 - 65951187 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01232361_m1
Size
Availability Made To Order
Catalog # 4351372
Price 4,430.00
Your Price
Online offer:
4430.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Twsg1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_023053.3 NP_075540.1 3 - 4 378 117
GenBank mRNA AF292033.1 - 317 117
AJ297390.1 - 3 - 4 328 117
AK033504.1 - 3 - 4 337 117
AK049646.1 - 3 - 4 372 117
AK168547.1 - 3 - 4 328 117
AK168633.1 - 3 - 4 337 117

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1700002418.mm.1

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Twsg1

Gene Ontology Categories:

Function(s) Process(es)

ossification
mesoderm formation
multicellular organism development
salivary gland morphogenesis
embryo development
tissue development
hemopoiesis
cell differentiation
BMP signaling pathway
positive regulation of BMP signaling pathway
negative regulation of BMP signaling pathway
forebrain development
camera-type eye development
negative regulation of osteoblast differentiation
protein binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1700022431.mm.1 NM_023053.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR1700022431.mm.1 NM_023053.3 Refseq
Affymetrix Mouse Clariom S Array PSR1700022431.mm.1 TC1700002418.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-9hc65:80/100.66.78.230:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0