This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01232856_g1
See other AWAT2 GE Assays ›
Gene Symbol:
Awat2
Gene Name:
acyl-CoA wax alcohol acyltransferase 2
Gene Aliases:
9430062J17Rik, ARAT, Dgat2l4, WS
Chromosome Location:
Chr.X: 100399336 - 100494221 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01232856_g1
Size
Availability Made To Order
Catalog # 4351372
Price 4,430.00
Your Price
Online offer:
4430.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Awat2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001290395.1 NP_001277324.1 3 - 4 622 68
NM_177746.4 NP_808414.2 2 - 3 252 68
GenBank mRNA AK034920.1 - 3 - 4 623 68
AY611031.1 - 2 - 3 227 68
AY611032.1 - 2 - 3 252 68
BC138085.1 - 2 - 3 220 8861104 68
BC138086.1 - 2 - 3 220 8861105 68

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0X00002703.mm.1

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Awat2

Panther Classification:

Molecular Function -

acyltransferase

Gene Ontology Categories:

Function(s) Process(es)

lipid metabolic process
wax biosynthetic process
molecular_function
transferase activity
transferase activity, transferring acyl groups
transferase activity, transferring acyl groups other than amino-acyl groups
long-chain-alcohol O-fatty-acyltransferase activity
retinol O-fatty-acyltransferase activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0X00016954.mm.1 NM_177746.4 Refseq
Affymetrix Mouse Clariom S Array PSR0X00016954.mm.1 TC0X00002703.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0X00016952.mm.1 NM_177746.4 Refseq
Affymetrix Mouse Clariom S Array PSR0X00016952.mm.1 TC0X00002703.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0X00016951.mm.1 NM_177746.4 Refseq
Affymetrix Mouse Clariom S Array PSR0X00016951.mm.1 TC0X00002703.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0X00016953.mm.1 NM_177746.4 Refseq
Affymetrix Mouse Clariom S Array PSR0X00016953.mm.1 TC0X00002703.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-2xklz:80/100.66.76.139:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0