This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01232942_m1
See other CRIM1 GE Assays ›
Gene Symbol:
Crim1
Gene Name:
cysteine rich transmembrane BMP regulator 1 (chordin like)
Gene Aliases:
AU015004
Chromosome Location:
Chr.17: 78200185 - 78376592 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01232942_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Crim1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_015800.3 NP_056615.1 13 - 14 2943 69
GenBank mRNA AF168680.1 - 13 - 14 2395 69
AK034889.1 - 1 - 2 171 69
AK053197.1 - 11 - 12 1886 69
BC100348.1 - 13 - 14 2728 30521148 69

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1700001178.mm.1

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Crim1

Gene Ontology Categories:

Function(s) Process(es)

regulation of cell growth
enzyme inhibitor activity
serine-type endopeptidase inhibitor activity
insulin-like growth factor binding
PDZ domain binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1700011189.mm.1 NM_015800.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR1700011189.mm.1 NM_015800.3 Refseq
Affymetrix Mouse Clariom S Array PSR1700011189.mm.1 TC1700001178.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-wt24k:80/100.66.72.70:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline