This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01232952_m1
See other CRIM1 GE Assays ›
Gene Symbol:
Crim1
Gene Name:
cysteine rich transmembrane BMP regulator 1 (chordin like)
Gene Aliases:
AU015004
Chromosome Location:
Chr.17: 78200185 - 78376592 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01232952_m1
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Crim1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_015800.3 NP_056615.1 7 - 8 1893 83
GenBank mRNA AF168680.1 - 7 - 8 1345 83
AK053197.1 - 5 - 6 836 83
BC100348.1 - 7 - 8 1678 30521148 83

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Crim1

Gene Ontology Categories:

Function(s) Process(es)

regulation of cell growth
enzyme inhibitor activity
serine-type endopeptidase inhibitor activity
insulin-like growth factor binding
PDZ domain binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-744bc48644-454p7:80/100.66.72.101:80.
git-commit: 747bde55a712f6f97bf7760408d445eefba4e16f
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.48.0-Offline