This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01232976_m1
See other PPM1B GE Assays ›
Gene Symbol:
Ppm1b
Gene Name:
protein phosphatase 1B, magnesium dependent, beta isoform
Gene Aliases:
PP2CB
Chromosome Location:
Chr.17: 84956741 - 85023992 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01232976_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Ppm1b
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001159498.1 NP_001152970.1 6 - 7 1267 89
GenBank mRNA AK077136.1 - 7 - 8 1340 89
BQ176513.1 - 1 - 2 89
D45859.1 - 5 - 6 463 89
D45861.1 - 6 - 7 490 89
U09218.1 - 6 - 7 1275 89

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1700001266.mm.1

Set Membership:

Amplicon length between 86 and 100 Protein Phosphatase 3' Most Mus musculus Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

protein dephosphorylation
N-terminal protein myristoylation
negative regulation of interferon-beta production
peptidyl-threonine dephosphorylation
negative regulation of NF-kappaB import into nucleus
negative regulation of I-kappaB kinase/NF-kappaB signaling
negative regulation of defense response to virus
magnesium ion binding
catalytic activity
phosphoprotein phosphatase activity
protein serine/threonine phosphatase activity
hydrolase activity
manganese ion binding
metal ion binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1700011716.mm.1 NM_001159498.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1700011716.mm.1 NM_001159498.1 Refseq
Affymetrix Mouse Clariom S Array PSR1700011716.mm.1 TC1700001266.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-55658555d6-fl825:80/100.66.75.127:80.
git-commit: ec8e7df5f8fec8d765bd419205f1b5046a016d37
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.41.0-Offline