This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01233075_g1
See other TXNRD1 GE Assays ›
Gene Symbol:
Txnrd1
Gene Name:
thioredoxin reductase 1
Gene Aliases:
TR, TR1, TrxR1
Chromosome Location:
Chr.10: 82833951 - 82897724 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01233075_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Txnrd1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001042513.1 NP_001035978.1 9 - 10 1102 86
NM_001042514.1 NP_001035979.1 8 - 9 1190 86
NM_001042523.1 NP_001035988.1 9 - 10 1319 86
NM_015762.2 NP_056577.2 8 - 9 995 86
GenBank mRNA AB027565.1 - 8 - 9 1022 86
AF333036.1 - 9 - 10 1319 86
AK011902.1 - 9 - 10 1103 86
AK146125.1 - 8 - 9 996 86
AK149625.1 - 9 - 10 1103 86
AK151927.1 - 9 - 10 1103 86
AK168356.1 - 9 - 10 1103 86
AK172383.1 - 7 - 8 3355 86
AK181192.1 - 2 - 3 204 86
AK183038.1 - 2 - 3 204 86
BC020274.1 - 8 - 9 957 86
BC037643.1 - 8 - 9 1275 5059831 86

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1000000933.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Txnrd1

Panther Classification:

Molecular Function -

reductase

Gene Ontology Categories:

Function(s) Process(es)

mesoderm formation
response to oxidative stress
gastrulation
cell proliferation
positive regulation of cell death
selenocysteine metabolic process
benzene-containing compound metabolic process
hydrogen peroxide catabolic process
cell redox homeostasis
protein tetramerization
oxidation-reduction process
NADPH oxidation
thioredoxin-disulfide reductase activity
NAD(P)H oxidase activity
oxidoreductase activity
oxidoreductase activity, acting on a sulfur group of donors, NAD(P) as acceptor
selenate reductase activity
protein homodimerization activity
mercury ion binding
flavin adenine dinucleotide binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-w64qk:80/100.66.78.26:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0