This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01233363_m1
See other EIF1AD GE Assays ›
Gene Symbol:
Eif1ad
Gene Name:
eukaryotic translation initiation factor 1A domain containing
Gene Aliases:
2010003J03Rik, AI131644
Chromosome Location:
Chr.19: 5366813 - 5371511 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01233363_m1
Size
Availability Made To Order
Catalog # 4351372
Price 4,430.00
Your Price
Online offer:
4430.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Eif1ad
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_027236.2 NP_081512.1 1 - 2 398 88
GenBank mRNA AK008070.1 - 1 - 2 208 88
AK031807.1 - 1 - 2 232 88
AK153194.1 - 1 - 2 229 88
AK158977.1 - 1 - 2 216 88
AK168102.1 - 1 - 2 202 88
AK168253.1 - 1 - 2 202 88
AK168598.1 - 1 - 2 225 88
BC024640.1 - 1 - 2 398 4038046 88

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the AK008070.1, AK153194.1, AK158977.1, AK168102.1, AK168253.1, AK168598.1, BC024640.1, NM_027236.2 transcripts.

Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus 1700 Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Eif1ad

Panther Classification:

Molecular Function -

translation factor translation initiation factor translational protein

Gene Ontology Categories:

Function(s) Process(es)

translational initiation
RNA binding
translation initiation factor activity

MicroArray Probes:

Affymetrix ABI

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1900000765.mm.1 NM_027236.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR1900000765.mm.1 NM_027236.2 Refseq
Affymetrix Mouse Clariom S Array PSR1900000765.mm.1 TC1900000062.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1900000768.mm.1 NM_027236.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR1900000768.mm.1 NM_027236.2 Refseq
Affymetrix Mouse Clariom S Array PSR1900000768.mm.1 TC1900000062.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1900000766.mm.1 NM_027236.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR1900000766.mm.1 NM_027236.2 Refseq
Affymetrix Mouse Clariom S Array PSR1900000766.mm.1 TC1900000062.mm.1 MTA TC
Array Name Probe IDs Targets Target Type
ABI Mouse Genome Survey Microarray 880049 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-2xklz:80/100.66.76.139:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0