This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01233367_m1
See other EIF1AD GE Assays ›
Gene Symbol:
Eif1ad
Gene Name:
eukaryotic translation initiation factor 1A domain containing
Gene Aliases:
2010003J03Rik, AI131644
Chromosome Location:
Chr.19: 5366813 - 5371511 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01233367_m1
Best Coverage
Size
Availability Inventoried
Catalog # 4331182
Price 2,694.00
Your Price
Online offer:
2694.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Eif1ad
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_027236.2 NP_081512.1 5 - 6 885 64
GenBank mRNA AK008070.1 - 5 - 6 695 64
AK031807.1 - 5 - 6 719 64
AK153194.1 - 5 - 6 716 64
AK158977.1 - 5 - 6 703 64
AK168102.1 - 5 - 6 689 64
AK168253.1 - 5 - 6 689 64
AK168598.1 - 5 - 6 712 64
BC024640.1 - 5 - 6 885 4038046 64

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1900000062.mm.1

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Eif1ad

Panther Classification:

Molecular Function -

translation factor translation initiation factor translational protein

Gene Ontology Categories:

Function(s) Process(es)

translational initiation
RNA binding
translation initiation factor activity

MicroArray Probes:

Agilent Affymetrix ABI Illumina

Array Name Probe IDs Targets Target Type
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31928286 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31928290 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31928289 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31928291 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31928287 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31928288 - -
Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2052081 BC024640.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2052081 AK153194.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2052081 AK158977.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2052081 AK168102.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2052081 AK168253.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P278154 BC024640.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P278154 AK158977.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P278154 AK031807.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P278154 NM_027236.2 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P278154 BC024640.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P278154 AK158977.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P278154 AK031807.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P278154 NM_027236.2 mouse_transcriptome
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 702078 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 694937 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 875678 - -
Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P152973 - -
SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P278154 BC024640.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P278154 AK158977.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P278154 AK031807.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P278154 NM_027236.2 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P278154 BC024640.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P278154 AK158977.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P278154 AK031807.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P278154 NM_027236.2 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2052081 BC024640.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2052081 AK153194.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2052081 AK158977.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2052081 AK168102.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2052081 AK168253.1 mouse_transcriptome
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31928301 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31928298 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31928295 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31928304 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31928296 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31928315 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31928322 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31928289 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31928311 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31928312 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31928318 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31928308 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31928290 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31928319 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31928313 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31928294 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31928303 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31928291 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31928286 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31928288 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31928306 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31928305 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31928316 - -
Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P152973 - -
Whole Mouse Genome G4122A A_52_P278154 BC024640.1 mouse_transcriptome
Whole Mouse Genome G4122A A_52_P278154 AK158977.1 mouse_transcriptome
Whole Mouse Genome G4122A A_52_P278154 AK031807.1 mouse_transcriptome
Whole Mouse Genome G4122A A_52_P278154 NM_027236.2 mouse_transcriptome
Whole Mouse Genome G4122A A_52_P278154 BC024640.1 mouse_transcriptome
Whole Mouse Genome G4122A A_52_P278154 AK158977.1 mouse_transcriptome
Whole Mouse Genome G4122A A_52_P278154 AK031807.1 mouse_transcriptome
Whole Mouse Genome G4122A A_52_P278154 NM_027236.2 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P474688 BC024640.1 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P474688 AK153194.1 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P474688 AK158977.1 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P474688 AK168102.1 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P474688 AK168253.1 mouse_transcriptome
Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P152973 - -
Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P14286748 - -
Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P14286746 - -
Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P14286751 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31928288 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31928287 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31928291 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31928289 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31928290 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31928286 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31928306 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31928372 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31928311 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31928290 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31928366 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31928357 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31928319 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31928308 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31928315 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31928380 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31928284 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31928354 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31928305 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31928294 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31928288 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31928378 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31928296 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31928301 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31928324 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31928374 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31928376 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31928355 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31928362 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31928364 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31928289 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31928316 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31928368 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31928298 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31928295 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31928313 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31928323 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31928359 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31928370 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31928329 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31928286 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31928327 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31928318 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P31928290 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P14324182 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P14324188 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P31928289 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P14314575 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P31928292 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P31928322 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P14416485 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P31928296 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P14141947 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P14216053 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P31928298 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P31928325 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P31928301 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P31928286 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P31928305 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P31928326 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P14271350 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P31928303 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P14401917 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P14401918 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P31928313 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P31928312 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P31928311 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P31928315 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P31928294 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P31928291 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P31928319 - -
Array Name Probe IDs Targets Target Type
Affymetrix Mouse Gene 1.0 ST Array 10460576 - -
Affymetrix Mouse Gene 1.0 ST Array 10460575 - -
Affymetrix Mouse Gene 1.0 ST Array 10460574 - -
Affymetrix Mouse Gene 1.0 ST Array 10460577 - -
Affymetrix Mouse Gene 1.0 ST Array 10460578 - -
Affymetrix Mouse Gene 1.0 ST Array 10460579 - -
Affymetrix HT MG-430 PM Array Plate 1428542_PM_at - -
Affymetrix HT MG-430 PM Array Plate 1455341_PM_at - -
Affymetrix Mouse Gene 1.1 ST Array 10460573 - -
Mouse Genome 430 2.0 Array 1455341_PM_at - -
Mouse Genome 430 2.0 Array 1455341_at - -
Mouse Genome 430 2.0 Array 1428542_PM_at - -
Mouse Genome 430 2.0 Array 1428542_at - -
Affymetrix GeneChip Mouse Genome 430 2.0 Array 69860_at - -
Affymetrix GeneChip Mouse Genome 430 2.0 Array 69860_PM_at - -
Affymetrix Mouse Clariom S Array PSR1900000778.mm.1 NM_027236.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR1900000778.mm.1 NM_027236.2 Refseq
Affymetrix Mouse Clariom S Array PSR1900000778.mm.1 TC1900000062.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1900000779.mm.1 NM_027236.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR1900000779.mm.1 NM_027236.2 Refseq
Affymetrix Mouse Clariom S Array PSR1900000779.mm.1 TC1900000062.mm.1 MTA TC
Mouse Expression Set 430 1428542_at - -
Mouse Expression Set 430 1428542_PM_at - -
Mouse Expression Set 430 1455341_PM_at - -
Mouse Expression Set 430 1455341_at - -
Affymetrix HT Mouse Genome 430B Array 1428542_PM_at - -
Affymetrix HT Mouse Genome 430B Array 1428542_at - -
Affymetrix HT Mouse Genome 430B Array 1455341_at - -
Affymetrix HT Mouse Genome 430B Array 1455341_PM_at - -
Array Name Probe IDs Targets Target Type
ABI Mouse Genome Survey Microarray 880049 - -
Array Name Probe IDs Targets Target Type
Illumina mouse-6 v1.1 expression beadchip ILMN_2683916 - -
Illumina mouse-6 v1.1 expression beadchip ILMN_1237895 - -
MouseRef-8 v2 Expression BeadChip ILMN_2968032 - -
Illumina MouseWG-6 v2.0 expression beadchip ILMN_2968032 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-zpsr4:80/100.66.78.116:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0