This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01233409_g1
See other MAGT1 GE Assays ›
Gene Symbol:
Magt1
Gene Name:
magnesium transporter 1
Gene Aliases:
2410001C15Rik, 2610529C04Rik, 2810482I07Rik, IAG2, IAP
Chromosome Location:
Chr.X: 105968084 - 106011899 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01233409_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Magt1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001190409.1 NP_001177338.1 6 - 7 894 79
NM_025952.4 NP_080228.4 6 - 7 894 79
GenBank mRNA AK010320.1 - 6 - 7 830 79
AK012185.1 - 6 - 7 771 79
AK013243.1 - 6 - 7 775 79
AK018623.1 - 6 - 7 797 79
AK136622.1 - 6 - 7 776 79
AK144696.1 - 6 - 7 773 79
BC003881.1 - 6 - 7 781 3496572 79
DQ000005.1 - 6 - 7 894 79

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0X00002825.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Magt1

Panther Classification:

Molecular Function -

glycosyltransferase

Gene Ontology Categories:

Function(s) Process(es)

transport
magnesium ion transport
cell redox homeostasis
cognition
cation transmembrane transport
magnesium ion transmembrane transporter activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0X00017938.mm.1 NM_001190409.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0X00017938.mm.1 NM_001190409.1 Refseq
Affymetrix Mouse Clariom S Array PSR0X00017938.mm.1 NM_025952.4.1 Refseq
Affymetrix Mouse Clariom S Array PSR0X00017938.mm.1 NM_025952.4 Refseq
Affymetrix Mouse Clariom S Array PSR0X00017938.mm.1 TC0X00002825.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-zvw94:80/100.66.78.114:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0