This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01233687_m1
See other STAU2 GE Assays ›
Gene Symbol:
Stau2
Gene Name:
staufen (RNA binding protein) homolog 2 (Drosophila)
Gene Aliases:
-
Chromosome Location:
Chr.1: 16228803 - 16520181 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01233687_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Stau2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001111272.1 NP_001104742.1 2 - 3 196 94
GenBank mRNA AF459100.1 - 2 - 3 146 94
AK171076.1 - 2 - 3 181 94
BC025118.1 - 2 - 3 151 3670067 94
CN665159.1 - 2 - 3 156 94

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the AF459100.1, BC025118.1, NM_001111272.1 transcripts.

Set Membership:

Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

transport
RNA binding
double-stranded RNA binding
poly(A) RNA binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0100016394.mm.1 NM_001111272.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0100016394.mm.1 NM_001111272.1 Refseq
Affymetrix Mouse Clariom S Array PSR0100016394.mm.1 NM_025303.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR0100016394.mm.1 NM_025303.3 Refseq
Affymetrix Mouse Clariom S Array PSR0100016394.mm.1 TC0100001999.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-8cb8z:80/100.66.72.77:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0