This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01233689_m1
See other STAU2 GE Assays ›
Gene Symbol:
Stau2
Gene Name:
staufen (RNA binding protein) homolog 2 (Drosophila)
Gene Aliases:
-
Chromosome Location:
Chr.1: 16228803 - 16520181 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01233689_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Stau2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001111272.1 NP_001104742.1 4 - 5 493 148
NM_025303.3 NP_079579.2 5 - 6 624 148
GenBank mRNA AF459099.1 - 5 - 6 574 148
AF459100.1 - 4 - 5 443 148
AK031102.1 - 5 - 6 552 148
BC010300.1 - 2 - 3 299 148
BC025118.1 - 4 - 5 448 3670067 148
BI987531.1 - 2 - 3 175 148
CN665159.1 - 4 - 5 453 148

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0100001999.mm.1

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Stau2

Gene Ontology Categories:

Function(s) Process(es)

transport
RNA binding
double-stranded RNA binding
poly(A) RNA binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0100016373.mm.1 NM_001111272.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0100016373.mm.1 NM_001111272.1 Refseq
Affymetrix Mouse Clariom S Array PSR0100016373.mm.1 NM_025303.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR0100016373.mm.1 NM_025303.3 Refseq
Affymetrix Mouse Clariom S Array PSR0100016373.mm.1 TC0100001999.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0100016374.mm.1 NM_001111272.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0100016374.mm.1 NM_001111272.1 Refseq
Affymetrix Mouse Clariom S Array PSR0100016374.mm.1 NM_025303.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR0100016374.mm.1 NM_025303.3 Refseq
Affymetrix Mouse Clariom S Array PSR0100016374.mm.1 TC0100001999.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-l4hk6:80/100.66.76.55:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0