This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01233807_g1
See other SOCS2 GE Assays ›
Gene Symbol:
Socs2
Gene Name:
suppressor of cytokine signaling 2
Gene Aliases:
8030460M17, AI527257, AW108012, CIS2, Cish2, D130043N08Rik, JAB, SOCS-2, SSI-2, hg
Chromosome Location:
Chr.10: 95383903 - 95417253 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01233807_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Socs2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_007706.4 NP_031732.1 1 - 2 236 96
GenBank mRNA AK137403.1 - 1 - 2 116 96
AK144810.1 - 1 - 2 177 96
AK160537.1 - 1 - 2 193 96
AK162591.1 - 1 - 2 201 96
BC106153.1 - 1 - 2 207 4989433 96
BY069559.1 - 1 - 2 236 96

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the AK137403.1,AK160537.1,AK162591.1,BC106153.1,NM_007706.4 transcripts.
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Apoptosis Mammalian Gene Collection Detect Genomic DNA Mus musculus 1700 Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Socs2

Panther Classification:

Molecular Function -

kinase modulator

Gene Ontology Categories:

Function(s) Process(es)

regulation of cell growth
negative regulation of protein kinase activity
lactation
negative regulation of signal transduction
cytokine-mediated signaling pathway
response to estradiol
cellular response to hormone stimulus
intracellular signal transduction
regulation of growth
regulation of multicellular organism growth
negative regulation of multicellular organism growth
positive regulation of neuron differentiation
negative regulation of JAK-STAT cascade
negative regulation of insulin receptor signaling pathway
growth hormone receptor signaling pathway
mammary gland alveolus development
protein kinase inhibitor activity
growth hormone receptor binding
insulin-like growth factor receptor binding
JAK pathway signal transduction adaptor activity

MicroArray Probes:

Affymetrix ABI

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1000020947.mm.1 NM_007706.4.1 Refseq
Affymetrix Mouse Clariom S Array PSR1000020947.mm.1 NM_007706.4 Refseq
Affymetrix Mouse Clariom S Array PSR1000020947.mm.1 TC1000002725.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1000020948.mm.1 NM_007706.4.1 Refseq
Affymetrix Mouse Clariom S Array PSR1000020948.mm.1 NM_007706.4 Refseq
Affymetrix Mouse Clariom S Array PSR1000020948.mm.1 TC1000002725.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1000020942.mm.1 NM_001168655.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1000020942.mm.1 NM_001168655.1 Refseq
Affymetrix Mouse Clariom S Array PSR1000020942.mm.1 NM_001168656.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1000020942.mm.1 NM_001168656.1 Refseq
Affymetrix Mouse Clariom S Array PSR1000020942.mm.1 NM_001168657.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1000020942.mm.1 NM_001168657.1 Refseq
Affymetrix Mouse Clariom S Array PSR1000020942.mm.1 NM_007706.4.1 Refseq
Affymetrix Mouse Clariom S Array PSR1000020942.mm.1 NM_007706.4 Refseq
Affymetrix Mouse Clariom S Array PSR1000020942.mm.1 TC1000002725.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1000020949.mm.1 NM_007706.4.1 Refseq
Affymetrix Mouse Clariom S Array PSR1000020949.mm.1 NM_007706.4 Refseq
Affymetrix Mouse Clariom S Array PSR1000020949.mm.1 TC1000002725.mm.1 MTA TC
Array Name Probe IDs Targets Target Type
ABI Mouse Genome Survey Microarray 895264 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-49tpj:80/100.66.77.222:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline