This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01234209_m1
See other CPSF6 GE Assays ›
Gene Symbol:
Cpsf6
Gene Name:
cleavage and polyadenylation specific factor 6
Gene Aliases:
4733401N12Rik, AI256641, CFIM, CFIM68, HPBRII-4, HPBRII-7
Chromosome Location:
Chr.10: 117344667 - 117376986 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01234209_m1
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Cpsf6
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001013391.2 NP_001013409.1 3 - 4 451 108
GenBank mRNA AK046856.1 - 3 - 4 407 108
AK048615.1 - 3 - 4 385 108
AK077684.1 - 3 - 4 452 108
AK168764.1 - 3 - 4 416 108
AK199961.1 - 2 - 3 198 108
BC068133.1 - 3 - 4 427 6813785 108

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1000002932.mm.1

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Cpsf6

Panther Classification:

Molecular Function -

RNA processing factor

Gene Ontology Categories:

Function(s) Process(es)

mRNA polyadenylation
mRNA processing
protein tetramerization
nucleotide binding
nucleic acid binding
RNA binding
mRNA binding
poly(A) RNA binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1000022178.mm.1 NM_001013391.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR1000022178.mm.1 NM_001013391.2 Refseq
Affymetrix Mouse Clariom S Array PSR1000022178.mm.1 TC1000002932.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-744bc48644-5wn4c:80/100.66.76.150:80.
git-commit: 747bde55a712f6f97bf7760408d445eefba4e16f
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.48.0-Offline