This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01234211_m1
See other CPSF6 GE Assays ›
Gene Symbol:
Cpsf6
Gene Name:
cleavage and polyadenylation specific factor 6
Gene Aliases:
4733401N12Rik, AI256641, CFIM, CFIM68, HPBRII-4, HPBRII-7
Chromosome Location:
Chr.10: 117344667 - 117376986 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01234211_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Cpsf6
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001013391.2 NP_001013409.1 5 - 6 771 62
GenBank mRNA AK046856.1 - 5 - 6 727 62
AK048615.1 - 5 - 6 705 62
AK049749.1 - 5 - 6 770 62
AK077684.1 - 5 - 6 773 62
AK168764.1 - 5 - 6 736 62
BC068133.1 - 5 - 6 747 6813785 62

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Cpsf6

Panther Classification:

Molecular Function -

RNA processing factor

Gene Ontology Categories:

Function(s) Process(es)

mRNA polyadenylation
mRNA processing
protein tetramerization
nucleotide binding
nucleic acid binding
RNA binding
mRNA binding
poly(A) RNA binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-mnfm6:80/100.66.78.37:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline