This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01234393_m1
See other SATL1 GE Assays ›
Gene Symbol:
Satl1
Gene Name:
spermidine/spermine N1-acetyl transferase-like 1
Gene Aliases:
4930404K22Rik
Chromosome Location:
Chr.X: 112383207 - 112406779 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01234393_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Satl1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_028655.1 NP_082931.1 5 - 6 2086 118
GenBank mRNA AK015086.1 - 5 - 6 1129 118
BC138009.1 - 5 - 6 2086 8861028 118
BC138011.1 - 5 - 6 2086 8861030 118

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0X00002869.mm.1

Set Membership:

Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Mus musculus 1700 Probe spans exons

Panther Classification:

Molecular Function -

acetyltransferase

Gene Ontology Categories:

Function(s) Process(es)

biological_process
molecular_function
dihydrolipoamide branched chain acyltransferase activity
sterol O-acyltransferase activity
N-acetyltransferase activity
O-acyltransferase activity
palmitoleoyl [acyl-carrier-protein]-dependent acyltransferase activity
carnitine O-acyltransferase activity
acetyltransferase activity
C-acyltransferase activity
palmitoyltransferase activity
N-acyltransferase activity
acylglycerol O-acyltransferase activity
serine O-acyltransferase activity
O-acetyltransferase activity
O-octanoyltransferase activity
octanoyltransferase activity
O-palmitoyltransferase activity
S-acyltransferase activity
S-acetyltransferase activity
S-malonyltransferase activity
malonyltransferase activity
C-palmitoyltransferase activity
transferase activity
transferase activity, transferring acyl groups
succinyltransferase activity
N-succinyltransferase activity
O-succinyltransferase activity
S-succinyltransferase activity
sinapoyltransferase activity
O-sinapoyltransferase activity
peptidyl-lysine N6-myristoyltransferase activity
peptidyl-lysine N6-palmitoyltransferase activity
benzoyl acetate-CoA thiolase activity
3-hydroxybutyryl-CoA thiolase activity
3-ketopimelyl-CoA thiolase activity
N-palmitoyltransferase activity
myristoyltransferase activity
acyl-CoA N-acyltransferase activity
protein-cysteine S-myristoyltransferase activity
protein-cysteine S-acyltransferase activity
dihydrolipoamide S-acyltransferase activity
glucosaminyl-phosphotidylinositol O-acyltransferase activity
ergosterol O-acyltransferase activity
lanosterol O-acyltransferase activity
naphthyl-2-oxomethyl-succinyl-CoA succinyl transferase activity
2,4,4-trimethyl-3-oxopentanoyl-CoA 2-C-propanoyl transferase activity
2-methylhexanoyl-CoA C-acetyltransferase activity
butyryl-CoA 2-C-propionyltransferase activity
2,6-dimethyl-5-methylene-3-oxo-heptanoyl-CoA C-acetyltransferase activity
L-2-aminoadipate N-acetyltransferase activity
UDP-3-O-[3-hydroxymyristoyl] glucosamine N-acyltransferase activity
keto acid formate lyase activity
Ras palmitoyltransferase activity
azetidine-2-carboxylic acid acetyltransferase activity
peptidyl-lysine N-acetyltransferase activity, acting on acetyl phosphate as donor
acetyl-CoA:L-lysine N6-acetyltransferase

MicroArray Probes:

Agilent Affymetrix ABI Illumina

Array Name Probe IDs Targets Target Type
Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2019207 AK015086.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2019207 NM_028655.1 mouse_transcriptome
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 743920 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 413076 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 43723 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 463085 AF313800.1 genbank_mrna_mouse
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 463085 AK032423.1 genbank_mrna_mouse
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 463085 AK034117.1 genbank_mrna_mouse
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 463085 AK157222.1 genbank_mrna_mouse
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 463085 AK160127.1 genbank_mrna_mouse
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 463085 AK171964.1 genbank_mrna_mouse
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 463085 BC028339.1 genbank_mrna_mouse
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 463085 BC038173.1 genbank_mrna_mouse
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 463085 BC050233.1 genbank_mrna_mouse
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 463085 BC060690.1 genbank_mrna_mouse
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 463085 BC080775.1 genbank_mrna_mouse
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 463085 BC086693.1 genbank_mrna_mouse
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 463085 BC118965.2 genbank_mrna_mouse
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 463085 BC118966.1 genbank_mrna_mouse
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 463085 NM_138681.3 refseq_mrna_mouse_NM
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 183410 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 918951 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 344081 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 570528 AK016540.1 genbank_mrna_mouse
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 570528 AK028927.1 genbank_mrna_mouse
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 570528 AK030928.1 genbank_mrna_mouse
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 570528 AK031527.1 genbank_mrna_mouse
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 570528 AK036454.1 genbank_mrna_mouse
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 570528 AK036465.1 genbank_mrna_mouse
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 570528 AK042544.1 genbank_mrna_mouse
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 570528 AK051372.1 genbank_mrna_mouse
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 570528 AK154166.1 genbank_mrna_mouse
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 570528 AK164148.1 genbank_mrna_mouse
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 570528 AK170232.1 genbank_mrna_mouse
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 570528 AK177699.1 genbank_mrna_mouse
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 570528 BC023094.1 genbank_mrna_mouse
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 570528 NM_144535.3 refseq_mrna_mouse_NM
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 15072 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 88316 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 871450 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 778948 - -
Agilent SurePrint G3 Mouse CGH Microarray 4x180K 25155 - -
Agilent SurePrint G3 Mouse CGH Microarray 4x180K 70187 - -
Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P136980 - -
Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P104478 - -
SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2019207 AK015086.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2019207 NM_028655.1 mouse_transcriptome
Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P136980 - -
Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P09394245 - -
Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P104478 - -
Whole Mouse Genome G4122A A_51_P288608 AK015086.1 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P288608 BC138009.1 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P288608 BC138011.1 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P288608 NM_028655.1 mouse_transcriptome
Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P136980 - -
Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P104478 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32603140 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32603127 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32603155 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32603116 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32603136 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32603135 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32603117 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32603114 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32603153 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32603125 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32603152 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32603112 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32603134 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32603150 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P14610704 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32603129 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32603115 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32603119 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32603131 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32603120 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32603138 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32603118 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32603123 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32603130 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P14610703 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32603133 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32603122 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P14788425 - -
Array Name Probe IDs Targets Target Type
Affymetrix Mouse Gene 1.0 ST Array 10606493 - -
Affymetrix Mouse Gene 1.0 ST Array 10606492 - -
Affymetrix Mouse Gene 1.0 ST Array 10606490 - -
Affymetrix Mouse Gene 1.0 ST Array 10606494 - -
Affymetrix Mouse Gene 1.0 ST Array 10606489 - -
Affymetrix Mouse Gene 1.0 ST Array 10606491 - -
Affymetrix HT MG-430 PM Array Plate 1431573_PM_at - -
Affymetrix Mouse Gene 1.1 ST Array 10606488 - -
Mouse Genome 430 2.0 Array 1431573_at - -
Mouse Genome 430 2.0 Array 1431573_PM_at - -
Affymetrix GeneChip Mouse Genome 430 2.0 Array 73809_at - -
Affymetrix GeneChip Mouse Genome 430 2.0 Array 73809_PM_at - -
Affymetrix Mouse Clariom S Array PSR0X00018269.mm.1 NM_028655.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0X00018269.mm.1 NM_028655.1 Refseq
Affymetrix Mouse Clariom S Array PSR0X00018269.mm.1 TC0X00002869.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0X00018270.mm.1 NM_028655.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0X00018270.mm.1 NM_028655.1 Refseq
Affymetrix Mouse Clariom S Array PSR0X00018270.mm.1 TC0X00002869.mm.1 MTA TC
Mouse Expression Set 430 1431573_PM_at - -
Mouse Expression Set 430 1431573_at - -
Affymetrix HT Mouse Genome 430B Array 1431573_PM_at - -
Affymetrix HT Mouse Genome 430B Array 1431573_at - -
Array Name Probe IDs Targets Target Type
ABI Mouse Genome Survey Microarray 422948 - -
Array Name Probe IDs Targets Target Type
Illumina mouseRef-8 v1.1 expression beadchip ILMN_1244241 - -
Illumina mouse-6 v1.1 expression beadchip ILMN_1244241 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-wf7gw:80/100.66.73.223:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0