Assay Details
| Gene Symbol: Camkv | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_145621.2 | NP_663596.1 | 3 - 4 | 364 | 67 | |
| GenBank mRNA | AK085143.1 | - | 370 | 67 | ||
| AK147323.1 | - | 3 - 4 | 352 | 67 | ||
| BC017634.1 | - | 3 - 4 | 364 | 4527857 | 67 | |
| BC022169.1 | - | 3 - 4 | 359 | 67 | ||
| BC086762.1 | - | 3 - 4 | 348 | 67 | ||
| BC111033.1 | - | 3 - 4 | 359 | 5368225 | 67 | |
More Information
Updated Mapping Information
Set Membership: |
|

Camkv
Genomic Map