This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01234738_g1
See other CAMKV GE Assays ›
Gene Symbol:
Camkv
Gene Name:
CaM kinase-like vesicle-associated
Gene Aliases:
1G5, BB074618, BC017634
Chromosome Location:
Chr.9: 107935851 - 107949691 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01234738_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Camkv
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_145621.2 NP_663596.1 5 - 6 584 83
GenBank mRNA AK085143.1 - 590 83
AK147323.1 - 5 - 6 572 83
BC017634.1 - 5 - 6 584 4527857 83
BC022169.1 - 4 - 5 793 83
BC086762.1 - 4 - 5 782 83
BC111033.1 - 5 - 6 579 5368225 83

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0900001385.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Protein Kinase Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Camkv

Panther Classification:

Molecular Function -

non-receptor serine/threonine protein kinase

Gene Ontology Categories:

Function(s) Process(es)

protein phosphorylation
protein kinase activity
calmodulin binding
ATP binding
transferase activity, transferring phosphorus-containing groups

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0900010935.mm.1 NM_145621.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0900010935.mm.1 NM_145621.2 Refseq
Affymetrix Mouse Clariom S Array PSR0900010935.mm.1 TC0900001385.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-jbr9k:80/100.66.74.50:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0