This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01234803_g1
See other ZEB1 GE Assays ›
Gene Symbol:
Zeb1
Gene Name:
zinc finger E-box binding homeobox 1
Gene Aliases:
3110032K11Rik, AREB6, BZP, MEB1, Nil2, TCF-8, Tcf18, Tcf8, Tw, ZEB, Zfhep, Zfhx1a, Zfx1a, Zfx1ha, [delta]EF1
Chromosome Location:
Chr.18: 5591860 - 5775468 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01234803_g1
Size
Availability Made To Order
Catalog # 4426961
Price 434.00
Your Price
Online offer:
434.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Zeb1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
GenBank mRNA AK041408.1 - 2 - 3 481 63

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream predicted gene 10125 Gm10125 ENSMUSG00000063087,
Zeb1os
-48.9kb Anti-sense Chr. 18 - 5491501 - 5592437 on NCBI Build GRCm38
Downstream predicted gene,
41662
Gm41662 371.4kb Anti-sense Chr. 18 - 6014253 - 6024013 on NCBI Build GRCm38

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 Detect Genomic DNA Mus musculus Probe spans exons

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-9dqhd:80/100.66.72.70:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline