This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01235065_m1
See other HAL GE Assays ›
Gene Symbol:
Hal
Gene Name:
histidine ammonia lyase
Gene Aliases:
Hsd, his, histidase
Chromosome Location:
Chr.10: 93488768 - 93516743 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01235065_m1
Size
Availability Made To Order
Catalog # 4351372
Price 4,430.00
Your Price
Online offer:
4430.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Hal
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_010401.3 NP_034531.1 17 - 18 2016 74
GenBank mRNA AK014518.1 - 17 - 18 2016 74
AK017397.1 - 2 - 3 190 74
AK028963.1 - 17 - 18 1723 74
AK081409.1 - 17 - 18 1719 74
BC057637.1 - 17 - 18 1693 4211545 74
BC157999.1 - 17 - 18 1998 9088147 74
L07645.1 - 17 - 18 1700 74

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1000001133.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Hal

Panther Classification:

Molecular Function -

lyase

Gene Ontology Categories:

Function(s) Process(es)

histidine metabolic process
histidine catabolic process
biosynthetic process
catalytic activity
histidine ammonia-lyase activity
lyase activity
ammonia-lyase activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1000008573.mm.1 NM_010401.4 Refseq
Affymetrix Mouse Clariom S Array PSR1000008573.mm.1 TC1000001133.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-2xklz:80/100.66.76.139:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0