This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01235497_m1
See other P4HTM GE Assays ›
Gene Symbol:
P4htm
Gene Name:
prolyl 4-hydroxylase, transmembrane (endoplasmic reticulum)
Gene Aliases:
4933406E20Rik, AI853847, BB128974, Ph-4
Chromosome Location:
Chr.9: 108578826 - 108597876 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01235497_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: P4htm
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_028944.3 NP_083220.3 6 - 7 1114 77
GenBank mRNA AK016685.1 - 6 - 7 1088 77
AK046783.1 - 6 - 7 1126 77
AK047714.1 - 6 - 7 1114 77
BC111871.1 - 6 - 7 1114 40057593 77

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0900003065.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene P4htm

Panther Classification:

Molecular Function -

protein modifying enzyme

Gene Ontology Categories:

Function(s) Process(es)

regulation of erythrocyte differentiation
oxidation-reduction process
sulfonate dioxygenase activity
iron ion binding
oxidoreductase activity
oxidoreductase activity, acting on paired donors, with incorporation or reduction of molecular oxygen
oxidoreductase activity, acting on paired donors, with incorporation or reduction of molecular oxygen, 2-oxoglutarate as one donor, and incorporation of one atom each of oxygen into both donors
2,4-dichlorophenoxyacetate alpha-ketoglutarate dioxygenase activity
procollagen-proline dioxygenase activity
L-ascorbic acid binding
hypophosphite dioxygenase activity
DNA-N1-methyladenine dioxygenase activity
metal ion binding
dioxygenase activity
C-19 gibberellin 2-beta-dioxygenase activity
C-20 gibberellin 2-beta-dioxygenase activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0900025820.mm.1 NM_028944.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR0900025820.mm.1 NM_028944.3 Refseq
Affymetrix Mouse Clariom S Array PSR0900025820.mm.1 TC0900003065.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0900025821.mm.1 NM_028944.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR0900025821.mm.1 NM_028944.3 Refseq
Affymetrix Mouse Clariom S Array PSR0900025821.mm.1 TC0900003065.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-744bc48644-5wn4c:80/100.66.76.150:80.
git-commit: 747bde55a712f6f97bf7760408d445eefba4e16f
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.48.0-Offline