This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01236157_g1
See other RPS6KA4 GE Assays ›
Gene Symbol:
Rps6ka4
Gene Name:
ribosomal protein S6 kinase, polypeptide 4
Gene Aliases:
1110069D02Rik, 90kDa, AI848992, Msk2, mMSK2
Chromosome Location:
Chr.19: 6829080 - 6841045 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01236157_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Rps6ka4
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_019924.2 NP_064308.2 5 - 6 653 65
GenBank mRNA AF074714.1 - 5 - 6 625 65
AK089467.1 - 5 - 6 644 65
AK154674.1 - 5 - 6 652 65
BC012964.1 - 5 - 6 663 4160873 65

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1900001017.mm.1

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Protein Kinase Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Rps6ka4

Panther Classification:

Molecular Function -

protein modifying enzyme

Gene Ontology Categories:

Function(s) Process(es)

regulation of transcription, DNA-templated
protein phosphorylation
inflammatory response
phosphorylation
positive regulation of histone phosphorylation
positive regulation of histone acetylation
intracellular signal transduction
histone H3-S10 phosphorylation
histone H3-S28 phosphorylation
positive regulation of transcription from RNA polymerase II promoter
interleukin-1-mediated signaling pathway
nucleotide binding
magnesium ion binding
protein kinase activity
protein serine/threonine kinase activity
protein binding
ATP binding
kinase activity
transferase activity
transferase activity, transferring phosphorus-containing groups
histone kinase activity (H3-S10 specific)
mitogen-activated protein kinase p38 binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1900009401.mm.1 NM_019924.2 Refseq
Affymetrix Mouse Clariom S Array PSR1900009401.mm.1 TC1900001017.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-mnfm6:80/100.66.78.37:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline