This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01236179_g1
See other GPR137 GE Assays ›
Gene Symbol:
Gpr137
Gene Name:
G protein-coupled receptor 137
Gene Aliases:
AI428855
Chromosome Location:
Chr.19: 6938062 - 6942450 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01236179_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Gpr137
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001177361.1 NP_001170832.1 2 - 3 662 92
NM_207220.2 NP_997103.1 2 - 3 765 92
GenBank mRNA AK039612.1 - 2 - 3 265 92
AK143450.1 - 2 - 3 663 92
BC048078.1 - 3 - 4 637 6308873 92

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1900001023.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 G-protein coupled receptor Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Gpr137

Gene Ontology Categories:

Function(s) Process(es)

biological_process
molecular_function

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1900009499.mm.1 NM_001177361.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1900009499.mm.1 NM_001177361.1 Refseq
Affymetrix Mouse Clariom S Array PSR1900009499.mm.1 NM_207220.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR1900009499.mm.1 NM_207220.2 Refseq
Affymetrix Mouse Clariom S Array PSR1900009499.mm.1 TC1900001023.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1900009505.mm.1 NM_001177360.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1900009505.mm.1 NM_001177360.1 Refseq
Affymetrix Mouse Clariom S Array PSR1900009505.mm.1 NM_207220.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR1900009505.mm.1 NM_207220.2 Refseq
Affymetrix Mouse Clariom S Array PSR1900009505.mm.1 TC1900001023.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1900009500.mm.1 NM_001177361.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1900009500.mm.1 NM_001177361.1 Refseq
Affymetrix Mouse Clariom S Array PSR1900009500.mm.1 NM_001177362.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1900009500.mm.1 NM_001177362.1 Refseq
Affymetrix Mouse Clariom S Array PSR1900009500.mm.1 NM_207220.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR1900009500.mm.1 NM_207220.2 Refseq
Affymetrix Mouse Clariom S Array PSR1900009500.mm.1 TC1900001023.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1900009502.mm.1 NM_001177360.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1900009502.mm.1 NM_001177360.1 Refseq
Affymetrix Mouse Clariom S Array PSR1900009502.mm.1 NM_207220.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR1900009502.mm.1 NM_207220.2 Refseq
Affymetrix Mouse Clariom S Array PSR1900009502.mm.1 TC1900001023.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-2xklz:80/100.66.76.139:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0