This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01236515_m1
See other CARNS1 GE Assays ›
Gene Symbol:
Carns1
Gene Name:
carnosine synthase 1
Gene Aliases:
AI790298, Atpgd1, mKIAA1394
Chromosome Location:
Chr.19: 4164324 - 4175479 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01236515_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Carns1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_134148.2 NP_598909.2 3 - 4 375 90
GenBank mRNA AK129347.1 - 3 - 4 350 90

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Carns1

Gene Ontology Categories:

Function(s) Process(es)

carnosine biosynthetic process
nucleotide binding
ATP binding
ligase activity
ATPase activity
metal ion binding
carnosine synthase activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1900007886.mm.1 NM_134148.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR1900007886.mm.1 NM_134148.2 Refseq
Affymetrix Mouse Clariom S Array PSR1900007886.mm.1 TC1900000906.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-49tpj:80/100.66.77.222:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline