This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01236928_g1
See other SAFB2 GE Assays ›
Gene Symbol:
Safb2
Gene Name:
scaffold attachment factor B2
Gene Aliases:
AA389433, AI255170, mKIAA0138
Chromosome Location:
Chr.17: 56560971 - 56584672 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01236928_g1
Size
Availability Made To Order
Catalog # 4351372
Price 4,430.00
Your Price
Online offer:
4430.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Safb2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001029979.2 NP_001025150.2 17 - 18 2659 105
GenBank mRNA BC027837.1 - 4 - 5 625 105
BC028827.1 - 11 - 12 1373 105
BC050855.1 - 17 - 18 2593 105
BC057039.1 - 15 - 16 1789 105

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1700002331.mm.1

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Safb2

Gene Ontology Categories:

Function(s) Process(es)

transcription, DNA-templated
regulation of transcription, DNA-templated
biological_process
nucleotide binding
nucleic acid binding
DNA binding
RNA binding
identical protein binding
poly(A) RNA binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1700021791.mm.1 NM_001029979.3 Refseq
Affymetrix Mouse Clariom S Array PSR1700021791.mm.1 TC1700002331.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1700021793.mm.1 NM_001029979.3 Refseq
Affymetrix Mouse Clariom S Array PSR1700021793.mm.1 TC1700002331.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1700021792.mm.1 NM_001029979.3 Refseq
Affymetrix Mouse Clariom S Array PSR1700021792.mm.1 TC1700002331.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-7dtsd:80/100.66.76.204:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0