This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01236931_m1
See other SAFB2 GE Assays ›
Gene Symbol:
Safb2
Gene Name:
scaffold attachment factor B2
Gene Aliases:
AA389433, AI255170, mKIAA0138
Chromosome Location:
Chr.17: 56560971 - 56584672 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01236931_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Safb2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001029979.2 NP_001025150.2 2 - 3 396 82
GenBank mRNA AK147798.1 - 2 - 3 336 82
AK161772.1 - 2 - 3 376 82
AK172898.1 - 2 - 3 372 82
BC022609.1 - 2 - 3 337 82
BC050855.1 - 2 - 3 330 82
BC057039.1 - 2 - 3 325 82
CN723671.1 - 2 - 3 398 82

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1700002331.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Safb2

Gene Ontology Categories:

Function(s) Process(es)

transcription, DNA-templated
regulation of transcription, DNA-templated
biological_process
nucleotide binding
nucleic acid binding
DNA binding
RNA binding
identical protein binding
poly(A) RNA binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1700021826.mm.1 NM_001029979.3 Refseq
Affymetrix Mouse Clariom S Array PSR1700021826.mm.1 TC1700002331.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-mvcqg:80/100.66.75.199:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0