This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01236959_m1
See other ADGRE1 GE Assays ›
Gene Symbol:
Adgre1
Gene Name:
adhesion G protein-coupled receptor E1
Gene Aliases:
DD7A5-7, EGF-TM7, Emr1, F4/80, Gpf480, Ly71, TM7LN3
Chromosome Location:
Chr.17: 57358651 - 57483529 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01236959_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Adgre1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_010130.4 NP_034260.1 5 - 6 563 73
GenBank mRNA AK133202.1 - 5 - 6 539 73
AK149898.1 - 5 - 6 549 73
AK151303.1 - 5 - 6 535 73
AK151682.1 - 5 - 6 555 73
AK155862.1 - 5 - 6 514 73
BC075688.1 - 5 - 6 535 30609706 73
U66888.1 - 5 - 6 550 73
U66889.1 - 5 - 6 550 73
X93328.1 - 5 - 6 559 73

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1700001034.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 G-protein coupled receptor Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Adgre1

Panther Classification:

Molecular Function -

G-protein coupled receptor

Gene Ontology Categories:

Function(s) Process(es)

signal transduction
cell surface receptor signaling pathway
G-protein coupled receptor signaling pathway
signal transducer activity
transmembrane signaling receptor activity
G-protein coupled receptor activity
calcium ion binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1700009970.mm.1 NM_010130.4.1 Refseq
Affymetrix Mouse Clariom S Array PSR1700009970.mm.1 NM_010130.4 Refseq
Affymetrix Mouse Clariom S Array PSR1700009970.mm.1 TC1700001034.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-zvjvc:80/100.66.72.70:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline