This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01239646_m1
See other CCT4 GE Assays ›
Gene Symbol:
Cct4
Gene Name:
chaperonin containing Tcp1, subunit 4 (delta)
Gene Aliases:
2610204B21Rik, A45, C78323, Cctd
Chromosome Location:
Chr.11: 22990593 - 23003336 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01239646_m1
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Cct4
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_009837.1 NP_033967.1 2 - 3 341 126
GenBank mRNA AK010634.1 - 2 - 3 309 126
AK146242.1 - 2 - 3 311 126
AK150601.1 - 2 - 3 288 126
AK152173.1 - 2 - 3 288 126
AK152665.1 - 2 - 3 309 126
AK163369.1 - 2 - 3 296 126
AK165229.1 - 2 - 3 309 126
AK167026.1 - 2 - 3 309 126
AK167262.1 - 2 - 3 278 126
AK167657.1 - 2 - 3 286 126
AK167846.1 - 2 - 3 286 126
AK169339.1 - 2 - 3 288 126
AK200879.1 - 1 - 2 53 126
AK206188.1 - 1 - 2 53 126
AK208716.1 - 1 - 2 53 125
BC054773.1 - 2 - 3 301 6807250 126
L25913.1 - 2 - 3 180 126
Z31554.1 - 2 - 3 341 126

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1100000234.mm.1

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Cct4

Panther Classification:

Molecular Function -

chaperonin

Gene Ontology Categories:

Function(s) Process(es)

protein folding
binding of sperm to zona pellucida
toxin transport
nucleotide binding
protein binding
ATP binding
poly(A) RNA binding
unfolded protein binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1100001874.mm.1 NM_009837.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1100001874.mm.1 NM_009837.1 Refseq
Affymetrix Mouse Clariom S Array PSR1100001874.mm.1 TC1100000234.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-l4hk6:80/100.66.76.55:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0