This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01239744_m1
See other EYA1 GE Assays ›
Gene Symbol:
Eya1
Gene Name:
eyes absent 1 homolog (Drosophila)
Gene Aliases:
bor
Chromosome Location:
Chr.1: 14168953 - 14310265 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01239744_m1
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Eya1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001252192.1 NP_001239121.1 11 - 12 1624 89
NM_010164.2 NP_034294.2 12 - 13 1740 89
GenBank mRNA AK042412.1 - 11 - 12 1547 89
AK153788.1 - 12 - 13 1746 89
AK162111.1 - 4 - 5 481 89
BC060260.1 - 12 - 13 1740 6848408 89
U61110.1 - 11 - 12 1297 89

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Eya1

Gene Ontology Categories:

Function(s) Process(es)

establishment of mitotic spindle orientation
metanephros development
ureteric bud development
branching involved in ureteric bud morphogenesis
outflow tract morphogenesis
DNA repair
double-strand break repair
transcription, DNA-templated
regulation of transcription, DNA-templated
protein dephosphorylation
cellular response to DNA damage stimulus
multicellular organism development
pattern specification process
mesodermal cell fate specification
organ morphogenesis
response to ionizing radiation
striated muscle tissue development
chromatin modification
histone dephosphorylation
protein sumoylation
cellular protein localization
establishment or maintenance of apical/basal cell polarity
peptidyl-tyrosine dephosphorylation
aorta morphogenesis
ear morphogenesis
inner ear morphogenesis
outer ear morphogenesis
middle ear morphogenesis
cell fate commitment
regulation of neuron differentiation
positive regulation of DNA repair
positive regulation of Notch signaling pathway
positive regulation of transcription, DNA-templated
positive regulation of transcription from RNA polymerase II promoter
neuron fate specification
embryonic skeletal system morphogenesis
semicircular canal morphogenesis
anatomical structure development
positive regulation of epithelial cell proliferation
pharyngeal system development
lung epithelial cell differentiation
otic vesicle development
otic vesicle morphogenesis
positive regulation of secondary heart field cardioblast proliferation
cochlea morphogenesis
negative regulation of extrinsic apoptotic signaling pathway in absence of ligand
RNA binding
phosphoprotein phosphatase activity
protein tyrosine phosphatase activity
protein binding
hydrolase activity
metal ion binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-rpjph:80/100.66.75.107:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0